Skip to Content
Merck
All Photos(1)

Key Documents

B8270

Sigma-Aldrich

Anti-Bovine Serum antibody produced in rabbit

whole antiserum

Synonym(s):

Anti-Serum, bovine

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

bovine

technique(s)

immunoelectrophoresis: suitable

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

bovine ... Alb(280717)

Looking for similar products? Visit Product Comparison Guide

General description

Bovine serum is a component extracted from bovine blood. It serves as a nutrient supplement in various in vitro cell cultures. It is also used in the culture of cumulus-oocyte complexes. Anti-bovine serum antibody can be used in differential staining to estimate inner cell mass and trophectoderm cell count. Rabbit anti- bovine serum antibody reacts specifically with normal bovine serum.

Immunogen

Pooled normal bovine serum.

Application

Anti-Bovine Serum antibody produced in rabbit has been used as a supplement in Dulbecco′s modified eagle medium for culturing inner cell mass (ICM) and cohesive trophectoderm (TE).
Anti-bovine serum antibody (10% (v/v) in Hepes-SOF) has been used in immunosurgery and immunoelectrophoresis.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Chen et al.
The Journal of biological chemistry, 267(17), 12380-12386 (1992-06-15)
Fetal bovine serum (FBS) has been widely used in the culture of cumulus-oocyte complexes, since it is believed that FBS stabilizes the expanding cumulus extracellular matrix. In this study, we have identified a factor in FBS that is responsible for
The Effect of Cryopreservation on the Mouse Embryos at Various-pronuclear Stages.
M. C. Park
Journal of Animal Science, 22(2), 174 -1180 (2009)
S M Hosseini et al.
BMC developmental biology, 15, 49-49 (2015-12-19)
This study describes the generation and analysis of the transcriptional profile of bovine inner cell mass (ICM) and trophectoderm (TE), obtained from in vivo developed embryos by using a bovine-embryo specific array (EmbryoGENE) containing 37,238 probes. A total of 4,689
Transcriptome profiling of bovine inner cell mass and trophectoderm derived from in vivo generated blastocysts
Hosseini, S. M. and others
BMC Developmental Biology, 15(1), 49-49 (2015)
N Gopichandran et al.
Reproduction (Cambridge, England), 126(3), 299-308 (2003-09-13)
The formation of a viable blastocyst is dependent upon the establishment of a correct inner cell mass (ICM):trophectoderm cell ratio but little is known about the metabolism of the two cell populations or about the composition of blastocoel fluid. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service