Skip to Content
Merck
All Photos(1)

Key Documents

B7686

Sigma-Aldrich

1(S),9(R)-(−)-Bicuculline methchloride

≥97% (HPLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H20NO6Cl
CAS Number:
Molecular Weight:
417.84
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

solubility

H2O: >10 mg/mL

storage temp.

2-8°C

SMILES string

[Cl-].C[N+]1(C)CCc2cc3OCOc3cc2[C@H]1[C@@H]4OC(=O)c5c6OCOc6ccc45

InChI

1S/C21H20NO6.ClH/c1-22(2)6-5-11-7-15-16(26-9-25-15)8-13(11)18(22)19-12-3-4-14-20(27-10-24-14)17(12)21(23)28-19;/h3-4,7-8,18-19H,5-6,9-10H2,1-2H3;1H/q+1;/p-1/t18-,19+;/m0./s1

InChI key

RLJKFAMYSYWMND-GRTNUQQKSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

1(S),9(R)-(−)-Bicuculline methchloride is a GABAA receptor antagonist, which blocks Ca2+-activated potassium (SK) channels.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the GABAA Receptors and Potassium Channels pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Pfleger et al.
Experimental brain research, 104(1), 81-88 (1995-01-01)
The influence of GABAA receptors on orientation selectivity of cat complex cells was tested by iontophoresis of the GABAA receptor blockers bicuculline and N-methyl-bicuculline while stimulating with drifting sinusoidal gratings. Reduction of orientation tuning was markedly less than reported in
H Qian et al.
Nature, 361(6408), 162-164 (1993-01-14)
gamma-Aminobutyric acid (GABA) is the main inhibitory neurotransmitter in the central nervous system. Two classes of GABA receptors (GABAA and GABAB) have been identified. GABAA receptors are ligand-gated chloride channels that are competitively antagonized by bicuculline, noncompetitively blocked by picrotoxin
O Paulsen et al.
The Journal of physiology, 496 ( Pt 3), 759-772 (1996-11-01)
1. Spontaneous non-NMDA glutamate receptor-mediated EPSCs were recorded with the whole-cell patch-clamp technique from twenty-six neurones in the dorsal lateral geniculate nucleus in thalamic slices from guinea-pig. 2. Amplitude distributions of the EPSCs were skewed towards larger values. The skewness
M Müller et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(1), 257-261 (1993-01-01)
The morphological and functional consequences of epileptic activity were investigated by applying the convulsants bicuculline and/or picrotoxin to mature rat hippocampal slice cultures. After 3 days, some cells in all hippocampal subfields showed signs of degeneration, including swollen somata, vacuolation
Y Katz et al.
Brain research, 646(2), 235-241 (1994-05-23)
We examined the interactions of D,L-laudanosine, a potentially epileptogenic metabolite of the neuromuscular relaxant atracurium besylate, with gamma-aminobutyric acid (GABA) and opioid binding sites, all of which have been implicated in seizure activity. Laudanosine was almost ineffective at [3H]muscimol binding

Articles

We offer many products related to GABAA receptors for your research needs.

We offer many products related to GABAA receptors for your research needs.

We offer many products related to GABAA receptors for your research needs.

We offer many products related to GABAA receptors for your research needs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service