Skip to Content
Merck
All Photos(1)

Key Documents

B7561

Sigma-Aldrich

1(S),9(R)-(−)-Bicuculline methbromide

≥98% (HPLC), solid

Synonym(s):

(5S)-5-[(6R)-6,8-Dihydro-8-oxofuro[3,4-e]-1,3-benzodioxol-6-yl]-5,6,7,8-tetrahydro-6,6-dimethyl-1,3-dioxolo[4,5-g]isoquinolinium bromide (1:1)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20BrNO6
CAS Number:
Molecular Weight:
462.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

H2O: >10 mg/mL

storage temp.

2-8°C

SMILES string

[Br-].C[N+]1(C)CCc2cc3OCOc3cc2[C@H]1[C@@H]4OC(=O)c5c6OCOc6ccc45

InChI

1S/C21H20NO6.BrH/c1-22(2)6-5-11-7-15-16(26-9-25-15)8-13(11)18(22)19-12-3-4-14-20(27-10-24-14)17(12)21(23)28-19;/h3-4,7-8,18-19H,5-6,9-10H2,1-2H3;1H/q+1;/p-1/t18-,19+;/m0./s1

InChI key

BWXCECYGGMGBHD-GRTNUQQKSA-M

Looking for similar products? Visit Product Comparison Guide

Application

1(S),9(R)-(-)-Bicuculline methbromide has been used:
  • as a γ-aminobutyric acid (GABAA) receptor A antagonist: in cortical neuron culture
  • in artificial cerebrospinal fluid (ACSF) formature dentate granule neurons
  • in hypothalamic explant cultures

Biochem/physiol Actions

1(S),9(R)-(−)-Bicuculline methbromide is a GABAA receptor antagonist, which blocks Ca2+-activated potassium (SK) channels. It is the water-soluble derivative of (+)-bicuculline.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Delphine Franssen et al.
Endocrinology, 157(5), 1740-1750 (2016-03-08)
Rat sexual maturation is preceded by a reduction of the interpulse interval (IPI) of GnRH neurosecretion. This work aims at studying disruption of that neuroendocrine event in females after early exposure to a very low dose of bisphenol A (BPA)
Amar Sahay et al.
Nature, 472(7344), 466-470 (2011-04-05)
Adult hippocampal neurogenesis is a unique form of neural circuit plasticity that results in the generation of new neurons in the dentate gyrus throughout life. Neurons that arise in adults (adult-born neurons) show heightened synaptic plasticity during their maturation and
Linda F Hayward et al.
Brain research, 974(1-2), 153-166 (2003-05-14)
The parabrachial nucleus (PBN) is located in the rostral dorsolateral pons and has been identified as a critical relay for cardiovascular responses (sympathoexcitation and baroreflex attenuation) evoked by the dorsal periaqueductal gray (PAG). We examined the pattern of c-Fos protein
Qing Hui Chen et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 285(5), R1231-R1239 (2003-07-26)
Blockade of GABA-A receptors in the hypothalamic paraventricular nucleus (PVN) has been repeatedly shown to increase arterial blood pressure (ABP), heart rate (HR), and sympathetic nerve activity (SNA), but the mechanism(s) that underlies this response has not been determined. Here
Christina Wenzek et al.
iScience, 27(8), 110547-110547 (2024-08-23)
The immune system has emerged as an important target of thyroid hormones (THs); however, the role of TH in T cells has so far remained elusive. In this study, we assessed the effect of TH receptor α (TRα) signaling on activation

Articles

We offer many products related to GABAA receptors for your research needs.

We offer many products related to GABAA receptors for your research needs.

We offer many products related to GABAA receptors for your research needs.

We offer many products related to GABAA receptors for your research needs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service