Skip to Content
Merck

Skip To

B5934

Barbital Buffer

Synonym(s):

Barbital buffer solution, Sodium barbital buffer solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12161703
NACRES:
NA.32
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

When reconstituted to 1 liter each vial produces a 1× running buffer containing 60 mM sodium barbital, pH 8.6.

Quality Segment

form

powder

drug control

Home Office Schedule 3; psychotrope (France); kontrollierte Droge in Deutschland; (Spain); Decreto Lei 15/93: Tabela IV (Portugal); Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet, kontrollierte Droge in Deutschland

Application

Barbital buffer has been used:
  • to cast 1% agarose gels and to run agarose gel electrophoresis[1]
  • to perform electrophoresis for lipoprotein-X (Lp-X) and lipoprotein separation[2]
  • to perform electrophoresis for lipoprotein analysis[3]

Biochem/physiol Actions

The most commonly used buffer for immunoelectrophoresis or immunodiffusion of protein on agarose gels or cellulose acetate. It is also used for separation of serum proteins, lipoproteins and isoenzymes.

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
9330293283
Quality Level

100

Quality Level

100

Quality Level

200

form

powder

form

liquid

form

liquid

drug control

Home Office Schedule 3; psychotrope (France); kontrollierte Droge in Deutschland; (Spain); Decreto Lei 15/93: Tabela IV (Portugal); Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet

drug control

-

drug control

-

description

When reconstituted to 1 liter each vial produces a 1× running buffer containing 60 mM sodium barbital, pH 8.6.

description

-

description

-


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 2 - STOT SE 3

target_organs

Nervous system, Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. Por favor, envíenme la composición exacta del Barbital Buffer ref. B5934

    1 answer
    1. This product contains free acid and sodium salt of Barbital. No other buffers or salts have been added. As noted in the 'PROPERTIS' section, when reconstituted to 1 liter each vial produces a 1X running buffer containing 60 mM sodium barbital, pH 8.6.

      Helpful?

  2. Can barbital buffer B5934-12VL be used for the CIE method for Aspergillus precipitin testing?

    1 answer
    1. Barbital buffer has been used for various purposes, including casting 1% agarose gels and running agarose gel electrophoresis, performing electrophoresis for lipoprotein-X (Lp-X) and lipoprotein separation, as well as for lipoprotein analysis. However, there is no information available for using B5934 in a CIE method for Aspergillus precipitin testing.

      Helpful?

Reviews

No rating value

Active Filters