Skip to Content
Merck
All Photos(1)

Key Documents

93283

Sigma-Aldrich

Tris-EDTA buffer solution

BioUltra, for molecular biology, pH 8.0

Synonym(s):

TE buffer, TE buffer solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41105319
eCl@ss:
32129211
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

grade

for molecular biology

Quality Level

product line

BioUltra

form

liquid

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

pH

8.0±0.2 (25 °C)

cation traces

Al: ≤1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.04
λ: 280 nm Amax: 0.01

application(s)

sample preparation

SMILES string

NC(CO)(CO)CO.OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C10H16N2O8.C4H11NO3/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;5-4(1-6,2-7)3-8/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);6-8H,1-3,5H2

InChI key

VLEIUWBSEKKKFX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris EDTA is a biological buffer in research. It is a combination of Tris base and EDTA, a metal chelator. Tris provides buffering capacity, maintaining a stable pH in biological samples, while EDTA helps prevent metal ion interference by chelating divalent metal ions. Tris EDTA is often used in molecular biology techniques, such as DNA and RNA sample preparation, electrophoresis, and enzymatic reactions. It helps maintain the stability and integrity of nucleic acids by inhibiting the activity of nucleases and metal-dependent enzymes.

Application

TE buffer is useful for general extraction, suspension, washing, and storage buffer and in the preparation of DNA or RNA. Commonly used for DNA sequencing, restriction enzyme digestion; ligation and polymerase chain reaction (PCR) applications.

Features and Benefits

  • BioUltra grade - suitable for many molecular biology applications
  • DNase, RNase, phosphatase, and protease-free
  • High purity and free of trace metals

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marina Ishii et al.
Applied biochemistry and biotechnology, 137-140(1-12), 555-571 (2008-05-15)
The thermal stability of recombinant green fluorescent protein (GFP) in sodium chloride (NaCl) solutions at different concentrations, pH, and temperatures was evaluated by assaying the loss of fluorescence intensity as a measure of denaturation. GFP, extracted from Escherichia coli cells
Quan-Ping Su et al.
Vaccine, 25(6), 970-975 (2006-10-24)
We have demonstrated that phage display Candida albicans (C. albicans) LKVIRK epitope was protective in systemically infected C57BL/6J mice. The different development from precursor Ths, Th1 or Th2, will result in a protective or nonprotective immune response. To compare the
A A Khan et al.
Journal of industrial microbiology & biotechnology, 20(2), 90-94 (1998-06-27)
A rapid and sensitive method for the detection of genetically engineered microorganisms in soil and sediments has been devised by in vitro amplification of the target DNAs by a polymerase chain reaction. A cloned catechol 2,3-dioxygenase gene located on the
Inbar Ben-Zvi et al.
ESC heart failure, 7(3), 835-843 (2020-04-08)
The aims of the study are to assess the levels of coronary sinus (CS) miRNAs of systolic heart failure (HF) patients in samples obtained during cardiac resynchronization therapy (CRT) device implantation and compare them to the peripheral systemic venous miRNA
Stuart W Tompson et al.
Bio-protocol, 7(10) (2017-08-02)
There are several in silico programs that endeavor to predict the functional impact of an individual's sequence variation at splice donor/acceptor sites, but experimental confirmation is problematic without a source of RNA from the individual that carries the variant. With

Questions

1–5 of 5 Questions  
  1. Is this buffer composed by 10mM Tris and 1 mM EDTA?

    1 answer
    1. This buffer is comprised of 10 mM Tris-HCl and 1 mM disodium EDTA, pH 8.0.

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  4. What is the storage condition for TE buffer BioUltra

    1 answer
    1. This product may be stored at room temperature.

      Helpful?

  5. What is the concentration of Tris and EDTA in this buffer?

    1 answer
    1. This buffer is 10 mM Tris-HCl, 1 mM disodium EDTA, pH 8.0.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service