Skip to Content
Merck
All Photos(1)

Key Documents

63249

Sigma-Aldrich

Benzalkonium chloride solution

≥50% (via Cl), 50% in H2O

Synonym(s):

Alkylbenzyldimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

concentration

≥50% (via Cl)
50% in H2O

SMILES string

[Cl-].[N+](CCCCCCCCCC)(Cc1ccccc1)(C)C

InChI

1S/C19H34N.ClH/c1-4-5-6-7-8-9-10-14-17-20(2,3)18-19-15-12-11-13-16-19;/h11-13,15-16H,4-10,14,17-18H2,1-3H3;1H/q+1;/p-1

InChI key

TTZLKXKJIMOHHG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Benzalkonium chloride is a quaternary ammonium salt in ocular drug products.

Application

Benzalkonium chloride (BAC) can be used as:       
  • A cationic surfactant in chemical polymerization reactions.       
  • A phase-transfer catalyst for the quantitative determination of sulfide in the whole blood sample by extractive alkylation technique using GC-MS.  

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A simple, optimized method for the determination of sulphide in whole blood by GC-MS as a marker of bowel fermentation processes
Hyvspler R, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 770(1-2), 255-259 (2002)
Structural effects of cationic surfactants in polyacrylamide-surfactant interaction: Micellisation/adsorption of polyacrylamide.
Niranjan, PS and Upadhyay, SK
Indian Journal of Chemistry (2014)
In situ gelling systems based on Pluronic F127/Pluronic F68 formulations for ocular drug delivery.
Al Khateb, Kosai et al.
International Journal of Pharmaceutics, 502(1-2), 70-79 (2016)
Effects of benzalkonium chloride on growth and survival of Chang conjunctival cells
De Saint Jean M, et al.
Investigative Ophthalmology & Visual Science, 40(3) (1999)
A wall-less poly (vinyl alcohol) cryogel flow phantom with accurate scattering properties for transcranial Doppler ultrasound propagation channels analysis.
Weir, Alexander J et al.
IEEE Conference Publication, 2709-2712 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service