Skip to Content
Merck
All Photos(1)

Key Documents

B3383

Sigma-Aldrich

Benzidine dihydrochloride

≥99% (titration)

Synonym(s):

4,4′-Diaminobiphenyl dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2 · 2HCl
CAS Number:
Molecular Weight:
257.16
Beilstein:
3914846
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99% (titration)

form

powder

SMILES string

Cl[H].Cl[H].Nc1ccc(cc1)-c2ccc(N)cc2

InChI

1S/C12H12N2.2ClH/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10;;/h1-8H,13-14H2;2*1H

InChI key

RUAXWVDEYJEWRY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Benzidine (4,4′-Diaminobiphenyl) is an environmental genotoxin that posses increased risks of cancer to exposed people. Benzidine may be used as a reference material in procedures that analyze for benzidine. Benzidine can be used in studies on its mechanisms of genotoxicity.

Biochem/physiol Actions

Tumor initiator in mammary glands of experimental animal.

Warning

Cancer suspect agent.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K C Morton et al.
Carcinogenesis, 2(8), 747-752 (1981-01-01)
To evaluate the role of metabolism in benzidine (BZ) carcinogenesis, BZ and 2 or its metabolites, N,N'-diacetylbenzidine and N-hydroxy-N,N'-diacetylbenzidine (NOHDABZ), were given by i.p. injection to female CD rats twice weekly for 4 weeks beginning at 30 days of age.
Jungsun Park et al.
American journal of industrial medicine, 52(8), 625-632 (2009-06-30)
Many corporations move their manufacturing facilities or technologies from developed to developing countries. Stringent regulations have made it costly for industries to operate in developed, industrialized countries. In addition, labor costs are high in these countries, and there is increasing
Jaroslaw Cendrowski et al.
eLife, 9 (2020-08-17)
Intracellular transport undergoes remodeling upon cell differentiation, which involves cell type-specific regulators. Bone morphogenetic protein 2-inducible kinase (BMP2K) has been potentially implicated in endocytosis and cell differentiation but its molecular functions remained unknown. We discovered that its longer (L) and
Yan Cao et al.
Stem cells and development, 24(22), 2660-2673 (2015-07-17)
Autophagy is required for hematopoietic stem cell multilineage differentiation, but the underlying mechanism is unknown. Using a conditional mouse model and human leukemia cells, we uncovered a mechanistic link between autophagy and hematopoietic stem cell differentiation. Loss of autophagy in
Jung Woo Lee et al.
Chemical communications (Cambridge, England), 48(3), 422-424 (2011-11-15)
We report a facile route to synthesize size tunable Fe(3)O(4) nanoparticles (NPs)-carbon nitride nanotube (CNNT) hybrids. These hybrids showing the water-soluble property are proven to exhibit ultra high peroxidase mimetic activity compared to those of pure NPs, where a colorless

Articles

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service