Skip to Content
Merck
All Photos(1)

Key Documents

B1522

SAFC

Basal Medium Eagle

With Earle′s salts and sodium bicarbonate, without ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

BME

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

product name

Basal Medium Eagle, With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

L-glutamine: no
NaHCO3: yes
Earle’s salts (5% CO2): yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Eagle′s Basal Medium (BME) is one of the most used synthetic cell culture media. The tissue culture association recommends that the term "Eagle′s Basal Medium" be used only to refer to the formula developed to support HeLa cells. BME has previously been used in monolayer culture studies to assess the growth response of normal (WI-38) and transformed (mouse and HeLa) cells. BME is the precursor of Dulbecco′s Modified Eagle′s medium (DME) and Eagle′s Minimum Essential Medium (MEM).

Application

Basal Medium Eagle has been used:
  • as a component in the culture medium for cochlear organotypic cultures and salicylate treatment
  • to maintain cerebellar granule neurons (CGNs) to determine if there is a system that regulates the distribution and/or group dynamics of axonal mitochondria
  • in the preparation of cortical slices, serum-free medium, and slice culture medium for slice overlay assay to study the influence of extracellular signals on neuronal development

Reconstitution

Supplement with 0.292 g/L L-glutamine.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nozomu Matsumoto et al.
Molecular and cellular neurosciences, 119, 103704-103704 (2022-02-09)
In the central nervous system (CNS), many neurons develop axonal arbors that are crucial for information processing. Previous studies have demonstrated that premature axons contain motile and stationary mitochondria, and their balance is important for axonal arborization. However, the mechanisms
Franck Polleux et al.
Science's STKE : signal transduction knowledge environment, 2002(136), pl9-pl9 (2002-06-13)
We have developed a technique that allows characterization and identification of extracellular signals that regulate various aspects of neuronal differentiation. In this in vitro assay, dissociated cells isolated from the developing cerebral wall are labeled and cultured over organotypic cortical
L Wei et al.
Neuroscience, 168(1), 288-299 (2010-03-20)
Aspirin, whose active ingredient is sodium salicylate, is the most widely used drug worldwide, but it is not recommended for children because it may cause Reye's syndrome. High doses of salicylate also induce temporary hearing loss and tinnitus; while these
Bethtrice Thompson et al.
PloS one, 11(2), e0148619-e0148619 (2016-02-20)
Bone continuously undergoes remodeling by a tightly regulated process that involves osteoblast differentiation from Mesenchymal Stem Cells (MSC). However, commitment of MSC to osteoblastic lineage is a poorly understood process. Chromatin organization functions as a molecular gatekeeper of DNA functions.
G M Seigel et al.
Molecular and cellular neurosciences, 85, 202-210 (2017-11-08)
Microglia are glial-immune cells that are essential for the function and survival of the central nervous system. Microglia not only protect neural tissues from immunological insults, but also play a critical role in neural development and repair. However, little is

Articles

Cell culture protocol for passaging and splitting suspension cell lines

Calcium is an ionically stable divalent cation with important beneficial and toxic properties in cell culture. It is a component of a wide range of cell culture media.

Calcium is an ionically stable divalent cation with important beneficial and toxic properties in cell culture. It is a component of a wide range of cell culture media.

Calcium is an ionically stable divalent cation with important beneficial and toxic properties in cell culture. It is a component of a wide range of cell culture media.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service