Skip to Content
Merck
All Photos(2)

Key Documents

B1310

Sigma-Aldrich

Anti-BACH1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-BRCA1-associated C-terminal helicase 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~150 kDa

species reactivity

human

technique(s)

immunoprecipitation (IP): 5-10 μg using RIPA extract (0.5 mg) of human HeLa cells
indirect immunofluorescence: 10-20 μg/mL using human MCF-7 cells
microarray: suitable
western blot: 0.5-1.0 μg/mL using whole extracts of MCF-7 human breast carcinoma cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BRIP1(83990)
mouse ... Brip1(237911)
rat ... Brip1(360588)

General description

BTB Domain And CNC Homolog 1 (BACH1) is expressed in numerous tissues. The protein includes a nuclear localization signal in its helical domain.

Immunogen

synthetic peptide corresponding to amino acid residues 1233-1249 of human BACH1 with N-terminal added cysteine, conjugated to KLH.

Application

Anti-BACH1 antibody produced in rabbit has been used in:
  • immunoblotting
  • immunohistochemistry
  • western blotting

Polyclonal anti-BACH1 antibody produced in rabbit is suitable for immunoprecipitation at a concentration of 5-10 μg by using RIPA extract (0.5 mg) of human HeLa cells and for indirect immunofluorescence at 10-20 μg/mL using human MCF-7 cells. The product is also suitable for western blot analysis at a working concentration of 0.5-1.0 μg/mL by using whole extracts of MCF-7 human breast carcinoma cells. Further, anti-BACH1 antibody has shown suitability for microarray analysis.

Biochem/physiol Actions

BACH1 interacts directly with the C-terminal BCRT repeats in Breast cancer type1 protein (BRCA1), a tumor suppressor nuclear phosphoprotein that facilitates the repair of double-stranded DNA breaks. Bach1 also functions as transcription repressor in transfection assays using fibroblast cells and as a transcriptional activator and repressor, respectively, in cultured erythroid cells. Additionally, it is also involved in the coordination of transcription activation and repression by Maf family members such as MafK. Bach1 (BTB and CNC homology 1, basic leucine zipper transcription factor 1) inhibits oxidative stress-inducible genes and is a negative regulator of oxidative stress-induced cellular senescence.
BTB Domain And CNC Homolog 1 (BACH1) interaction between breast cancer type 1 (BRCA1) in vivo appears to be dependent on phosphorylation of Ser990 in the latter during the G2/M phase of the cell cycle. BACH1 mutations were found in patients with early-onset breast cancer. It displays a punctate nuclear staining pattern similar to that observed for BRCA1.

Target description

BACH1, BRCA1 interacting protein C-terminal helicase 1, encodes a protein that is a member of the RecQ DEAH helicase family and interacts with the BRCT repeats of breast cancer, type 1 (BRCA1). The bound complex is important in the normal double-strand br

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chun-Peng Zhao et al.
Biomedical reports, 5(5), 585-588 (2016-11-25)
In a previous study, the suppressor of IKBKE 1 expression level was confirmed to be higher in vincristine (VCR)-resistant HCT-8 (HCT-8/V) colon cancer cells than in non-VCR-resistant HCT-8 cells. In the current study, IKBKE 1 expression in VCR-resistant colon cancer
David W Clark et al.
Oncotarget, 6(30), 28816-28832 (2015-09-05)
Fanconi anemia (FA) is a rare genome instability syndrome with progressive bone marrow failure and cancer susceptibility. FANCJ is one of 17 genes mutated in FA-patients, comprises a DNA helicase that is vital for properly maintaining genomic stability and is
Zhongmin Han et al.
Journal of cancer research and therapeutics, 15(1), 38-41 (2019-03-19)
This study was aimed to investigate the relationship between the expression of drebrin (DBN1) gene and resistance in colon cancer to reveal the mechanism of tumor drug resistance and provide a basis for the reversal of this drug resistance in
Overexpression of suppressor of IKBKE 1 is associated with vincristine resistance in colon cancer cells
Zhao CP, et al.
Biomedical Reports, 5(5), 585-588 (2016)
Jia-Min Zhang et al.
Molecular cell, 81(5), 1027-1042 (2021-01-17)
Alternative lengthening of telomeres (ALT) is mediated by break-induced replication (BIR), but how BIR is regulated at telomeres is poorly understood. Here, we show that telomeric BIR is a self-perpetuating process. By tethering PML-IV to telomeres, we induced telomere clustering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service