17095
Wang resin
extent of labeling: 1.0 mmol/g loading
Synonym(s):
p-Alkoxy-benzyl alcohol polymer-bound, p-Alkoxybenzyl alcohol resin, [4-(Hydroxymethyl)phenoxymethyl]polystyrene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
solid
Quality Level
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
extent of labeling
1.0 mmol/g loading
matrix
crosslinked with 1% DVB
particle size
75-100 mesh
application(s)
peptide synthesis
functional group
alcohol
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Sara Ashrafi et al.
Water environment research : a research publication of the Water Environment Federation, 92(7), 969-974 (2020-01-07)
In this study, CoFe2 O4 nanoparticles supported on multi-walled carbon nanotubes (MWCNTs), as novel peroxymonosulfate (PMS) activator, were synthesized for degradation of Reactive Black 5 (RB5). The results showed that the maximum removal efficiencies of RB5 (100%), chemical oxygen demand
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service