Skip to Content
Merck
All Photos(1)

Documents

APOAC

Sigma-Aldrich

Annexin V-Cy3 Apoptosis Detection Kit

Synonym(s):

Red fluorescent Annexin V apoptosis assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.32

usage

sufficient for 200 tests

Quality Level

packaging

pkg of 1 kit

technique(s)

flow cytometry: suitable

application(s)

cell analysis
detection

detection method

fluorometric

shipped in

wet ice

storage temp.

2-8°C

Features and Benefits

Detects apoptosis earlier in the process than DNA-based assays such as TUNEL
  • Rapid labeling of cells. Cell staining takes only 10 minutes.
  • No cell fixation or processing required, reducing the detection time and allowing the cells to be used for further study.
  • 6-Carboxyfluorescein diacetate secondary dye is included with the kit to differentiate apoptotic cells from viable and necrotic cells.

Principle

Sigma′s Annexin V-Cy3 kit allows detection of annexin V bound to apoptotic cells by fluorescence microscopy. The Annexin V-Cy3 kit uses the dye Cy3.18 as the fluorochrome conjugated with annexin V. By microscopy, Cy3.18 fluoresces more brightly than the FITC conjugate. The kit includes the non-fluorescent compound 6-carboxyfluorescein diacetate (6-CFDA), which enters the cell and is hydrolyzed by the esterases present in living cells to the fluorescent compound 6-carboxyfluorescein, indicating that the cells are viable. This combination allows the differentiation among early apoptotic cells (annexin V positive, 6-CFDA positive), necrotic cells (annexin V positive, 6-CFDA negative), and viable cells (annexin V negative, 6-CFDA positive).

Legal Information

Cy3 is a trademark of Cytiva

Kit Components Only

Product No.
Description

  • Annexin V Cy3.18 Conjugate from human placenta, buffered aqueous solution 10 μg

Kit Components Also Available Separately

Product No.
Description
SDS

  • C50416-Carboxyfluorescein diacetate 10 mgSDS

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kasturi Muthoosamy et al.
Scientific reports, 6, 32808-32808 (2016-09-07)
Metastasis of lung carcinoma to breast and vice versa accounts for one of the vast majority of cancer deaths. Synergistic treatments are proven to be the effective method to inhibit malignant cell proliferation. It is highly advantageous to use the
Christo Paul Preethy et al.
Journal of pharmacology & pharmacotherapeutics, 3(1), 26-34 (2012-03-01)
To find the efficacy of serial extracts of Anisomeles malabarica in inhibiting proliferation of and inducing apoptosis in human cervical cancer cells, SiHa and ME 180, that are HPV 16-positive. The whole plant was extracted in n-hexane, chloroform, ethyl acetate
Rajeev Kumar Pandey et al.
PloS one, 6(2), e17093-e17093 (2011-02-25)
Mycobacterium indicus pranii (MIP), also known as Mw, is a saprophytic, non-pathogenic strain of Mycobacterium and is commercially available as a heat-killed vaccine for leprosy and recently tuberculosis (TB) as part of MDT. In this study we provide evidence that
Javier Egea et al.
Journal of neurochemistry, 102(6), 1842-1852 (2007-06-02)
Activation of neuronal nicotinic acetylcholine receptors (nAChR) provides neuroprotection against different toxic stimuli that often lead to overproduction of reactive oxygen species (ROS) and cell death. ROS production has been related with disease progression in several neurodegenerative pathologies such as
Anvarbatcha Riyasdeen et al.
Evidence-based complementary and alternative medicine : eCAM, 2012, 136527-136527 (2011-11-25)
Rasagenthi Mezhugu (RGM) is a herbomineral formulation in the Siddha system of traditional medicine and is prescribed in the southern parts of India as a remedy for all kinds of cancers. However, scientific evidence for its therapeutic efficacy in cervical

Articles

Cellular apoptosis assays to detect programmed cell death using Annexin V, Caspase and TUNEL DNA fragmentation assays.

Cellular apoptosis assays to detect programmed cell death using Annexin V, Caspase and TUNEL DNA fragmentation assays.

Cellular apoptosis assays to detect programmed cell death using Annexin V, Caspase and TUNEL DNA fragmentation assays.

Cellular apoptosis assays to detect programmed cell death using Annexin V, Caspase and TUNEL DNA fragmentation assays.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service