Skip to Content
Merck
All Photos(1)

Documents

A9941

Sigma-Aldrich

2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt

chromogenic, tablet

Synonym(s):

AzBTS-(NH4)2, Diammonium 2,2′-azino-bis(3-ethylbenzothiazoline-6-sulfonate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24N6O6S4
CAS Number:
Molecular Weight:
548.68
Beilstein:
7329461
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.21

product name

2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, tablet, 10 mg substrate per tablet

Quality Level

form

tablet

solubility

deionized water: 100 mL

storage temp.

room temp

SMILES string

[NH4+].[NH4+].CCN1C(\Sc2cc(ccc12)S([O-])(=O)=O)=N\N=C3/Sc4cc(ccc4N3CC)S([O-])(=O)=O

InChI

1S/C18H18N4O6S4.2H3N/c1-3-21-13-7-5-11(31(23,24)25)9-15(13)29-17(21)19-20-18-22(4-2)14-8-6-12(32(26,27)28)10-16(14)30-18;;/h5-10H,3-4H2,1-2H3,(H,23,24,25)(H,26,27,28);2*1H3/b19-17-,20-18-;;

InChI key

OHDRQQURAXLVGJ-AXMZSLBLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

ABTS mainly used in ELISA (enzyme-linked immunosorbent assay) procedures.
ABTS, 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid), is a chromogenic substrate used:
  • For a reaction with laccase or bilirubin oxidase
  • For a reaction with peroxidase enzymes: (like HRP for example). ABTS in combination with hydrogen peroxide
A peroxidase reaction of ABTS in the presence of hydrogen peroxide produces a green soluble end product which can be read spectrophotometrically at 405nm. The reaction may be stopped with 1% sodium dodecyl sulfate (SDS).
ABTS, 2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt is light sensitive, and water soluble chemical compound

Application

  • 2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, ABTS, has been used as a substrate in ELISA (enzyme-linked immunosorbent assay).†
  • ABTS is also used to measure glucose concentrations in different solutions including blood serum.
  • The food industry uses ABTS to measure the antioxidant capacities within different foods.
  • Since it is less readily oxidized than TMB and OPD substrates, ABTS is less sensitive for ELISA applications. Thus ABTS has some advantageous over TMB and OPD in cases of large background results
  • It is recommended for ELISA (microwell) procedures, not recommended for membrane applications.
2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is green in color and can be read spectrophotometrically at 405 nm. The reaction may be stopped with 1% sodium dodecyl sulfate (SDS). Recommended for ELISA (microwell) procedures, not recommended for membrane applications.

Reconstitution

Dissolve 1 tablet in 100 ml of 0.05 M phosphate-citrate buffer, pH 5.0 (Tablets available as Sigma Product No. P4809). Add 25 μl of fresh 30% hydrogen peroxide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vignesh Narayanaswamy et al.
Breastfeeding medicine : the official journal of the Academy of Breastfeeding Medicine, 16(12), 987-994 (2021-08-13)
Objective: To evaluate the immune response to severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) in colostrum from women who tested positive for the virus. Methods: Between March and September 2020 we obtained bilateral colostrum samples collected on spot cards within
B Porstmann et al.
Journal of clinical chemistry and clinical biochemistry. Zeitschrift fur klinische Chemie und klinische Biochemie, 19(7), 435-439 (1981-07-01)
o-Phenylenediamine, 2,2'-azino-di(3-ethylbenzthiazoline sulphonic acid-6) (ABTS), o-dianisidine and 4-aminoantipyrine were compared as chromogens for the determination of horseradish peroxidase. Highest sensitivity in the determination of horseradish peroxidase-IgG conjugates in dissolved form was obtained with o-phenylenediamine. When these conjugates were used in
Development of a feline proinsulin immunoradiometric assay and a feline proinsulin enzyme-linked immunosorbent assay (ELISA): A novel application to examine beta cell function in cats
Kley S, et al.
Domestic Animal Endocrinology, 311-318 (2008)
H Gallati
Journal of clinical chemistry and clinical biochemistry. Zeitschrift fur klinische Chemie und klinische Biochemie, 17(1), 1-7 (1979-01-01)
The reaction of the two substrates hydrogen peroxide and ABTS with horseradish peroxidase was studied kinetically. Enzyme activity was determined as a function of substrate concentration and pH. Michaelis constants were determined for the two substrates at various pH values.
Collaborative study on a guinea pig serological method for the assay of acellular pertussis vaccines
Winsnes R, et al.
Pharmeuropa Bio & Scientific Notes, 19-32 (2009)

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Protocols

To measure alcohol oxidase activity, this assay uses 2,2′-azino-bis-(3-ethylbenzothiazoline-6-sulfonic acid) and a continuous spectrophotometric rate determination at 405 nm.

To measure alcohol oxidase activity, this assay uses 2,2′-azino-bis-(3-ethylbenzothiazoline-6-sulfonic acid) and a continuous spectrophotometric rate determination at 405 nm.

To measure alcohol oxidase activity, this assay uses 2,2′-azino-bis-(3-ethylbenzothiazoline-6-sulfonic acid) and a continuous spectrophotometric rate determination at 405 nm.

To measure alcohol oxidase activity, this assay uses 2,2′-azino-bis-(3-ethylbenzothiazoline-6-sulfonic acid) and a continuous spectrophotometric rate determination at 405 nm.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service