Skip to Content
Merck
All Photos(2)

Key Documents

11557

Sigma-Aldrich

2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt

chromogenic, ≥98% (HPLC), powder or crystals

Synonym(s):

AzBTS-(NH4)2, Diammonium 2,2′-azino-bis(3-ethylbenzothiazoline-6-sulfonate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24N6O6S4
CAS Number:
Molecular Weight:
548.68
Beilstein:
7329461
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83
Pricing and availability is not currently available.

Product Name

2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder or crystals

impurities

≤6.0% water

solubility

H2O: 10 mg/mL, slightly hazy to clear

storage temp.

2-8°C

SMILES string

[NH4+].[NH4+].CCN1C(\Sc2cc(ccc12)S([O-])(=O)=O)=N\N=C3/Sc4cc(ccc4N3CC)S([O-])(=O)=O

InChI

1S/C18H18N4O6S4.2H3N/c1-3-21-13-7-5-11(31(23,24)25)9-15(13)29-17(21)19-20-18-22(4-2)14-8-6-12(32(26,27)28)10-16(14)30-18;;/h5-10H,3-4H2,1-2H3,(H,23,24,25)(H,26,27,28);2*1H3/b19-17-,20-18-;;

InChI key

OHDRQQURAXLVGJ-AXMZSLBLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt has been used:
  • in the radical scavenging assay to test the antioxidant property of Pinus densiflora bark (PDB) extracts[1]
  • to test the antioxidant capacity (AOC) of Aloe vera samples[2]
  • as a substrate for streptavidin-horse radish peroxidase (HRPO) in enzyme-linked immunosorbent assay (ELISA)[3]

2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is green in color and can be read spectrophotometrically at 405 nm. The reaction may be stopped with 1% sodium dodecyl sulfate (SDS). Recommended for ELISA (microwell) procedures, not recommended for membrane applications.

Other Notes

Chromogenic substrate for peroxidasecitation; The ABTS-H2O2-peroxidase system in EIAcitationcitation
Chromogenic substrate for peroxidase[4]; The ABTS-H2O2-peroxidase system in EIA[5][6]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antioxidant and cytotoxicological effects of Aloe vera food supplements
Lopez Z, et al.
Journal of Food Quality, 2017 (2017)
A Szutowicz et al.
Analytical biochemistry, 138(1), 86-94 (1984-04-01)
Monoamine oxidase is assayed in tissue by a colorimetric reaction using horse radish peroxidase and 2,2'-azinodi(ethylbenzthiazoline-6-sulfonic acid to measure H2O2 formed during oxidation of amines. The method has a coefficient of variation of approximately 2.5% and provides results comparable with
Thamizhiniyan Venkatesan et al.
BioMed research international, 2019, 3520675-3520675 (2019-08-31)
It is well established that various extraction factors, including the method, temperature, time, and solvent system, significantly influence the antioxidant quality of plant-derived products. Previously, we observed that extraction of Pinus densiflora bark (PDB) by the most common traditional Soxhlet
Enzyme immunoassay ELISA and EMIT.
E Engvall
Methods in enzymology, 70(A), 419-439 (1980-01-01)
Jianwei Dong et al.
Journal of bioscience and bioengineering, 118(4), 396-399 (2014-04-20)
The tubers of Bletilla formosana were fermented with eight plant pathogen fungi, respectively, and antioxidant activities and total phenolic content (TPC) of the crude extracts of fermented products and non-fermented products were investigated. The antioxidant activities were evaluated in three

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service