Skip to Content
Merck

Skip To

A9799

Agar

High gel strength, suitable for plant cell culture

Synonym(s):

Agar-agar, Gum agar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(C12H18O9)n
CAS Number:
UNSPSC Code:
50405901
NACRES:
NA.21
EC Number:
232-658-1
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

algae (Rhodophyceae)

Quality Segment

form

powder

technique(s)

cell culture | plant: suitable

transition temp

transition temp 32-38 °C (1.5%)

gel strength

>1000 g/cm2

application(s)

agriculture

storage temp.

room temp

SMILES string

O1[C@@H]([C@@H]([C@@H]([C@H](C1OC2[C@H]3OCC2OC([C@H]3O)C)O)OC)O)CO

InChI

1S/C14H24O9/c1-5-8(16)13-11(7(21-5)4-20-13)23-14-10(18)12(19-2)9(17)6(3-15)22-14/h5-18H,3-4H2,1-2H3/t5?,6-,7?,8-,9+,10-,11?,12+,13+,14?/m1/s1

InChI key

GYYDPBCUIJTIBM-DYOGSRDZSA-N

General description

Agar is a natural polymer extracted from red alga Rhodophyceae, primarily, consisting of alternative D- and L-galactose units. This type of high gel strength agar is used when a firmer gel is required and mostly suitable for plant cell culture.
Agar is a polysaccharide complex obtained through bleaching and hot water extraction of agarocytes from the red alga Rhodophyceae, found in the Pacific and Indian Oceans and the sea of Japan. The genera Gelidium, Acanthopeltis, Ceramium, Pterocladia and Gracilaria predominate in agar production. Agar is composed of about 70% agarose and 30% agaropectin.

Application

Agar has been used:
  • in the preparation of laboratory diet to maintain the culture of D. melanogaster wild-type larva[1]
  • as a high gel strength agar in the seed culture media to grow Arabidopsis thaliana mutant seeds[2]
  • to construct a tissue-mimicking phantom to test the imaging performance of an ultrasound transducer[3]

Agar has been used:
  • as a component of the fruit fly diet[1]
  • as a component of saline-agar for the approximation of gray matter from brain[4]
  • in in vitro root nodulation assay of Glycine max seeds

Biochem/physiol Actions

Agar with a higher percentage of anhydrous units in the backbone of their chemical composition provides superior mechanical stiffness and strength to the pristine hydrogel.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A7921A480005039
form

powder

form

powder

form

powder

form

powder

gel strength

>1000 g/cm2

gel strength

-

gel strength

-

gel strength

>800 g/cm2 (1.5% gel, Nikkan, after autoclaving)

technique(s)

cell culture | plant: suitable

technique(s)

cell culture | plant: suitable

technique(s)

cell culture | plant: suitable

technique(s)

-

biological source

algae (Rhodophyceae)

biological source

algae (Rhodophyceae)

biological source

algae (Rhodophyceae)

biological source

-

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100


Still not finding the right product?

Explore all of our products under Agar


Storage Class

11 - Combustible Solids

wgk

WGK 1

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters