Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Agar
Synonym(s):
Agar-agar, Gum agar
Linear Formula:
(C
12
H
18
O
9
)
n
CAS Number:
9002-18-0
EC Number:
232-658-1
Products
Product Category
carbohydrates (8)
gelling agents (6)
binders (1)
polymers (1)
thickeners (1)
Brand
Millipore (7)
SAFC (1)
Sigma-Aldrich (8)
Biological Source
Feature
Melting Point (°C)
pH Value
Physical Form
Research Area
Quality Segment
Application
Reference Material Type
Sterility
Technique
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
A1296
Agar,
microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Expand
View Pricing
A7921
Agar,
suitable for plant cell culture, powder
Expand
View Pricing
05040
Agar,
powder, suitable for microbiology
Expand
View Pricing
A6686
Agar,
Bacteriological, microbiology tested, suitable for plant cell culture, granular
Expand
View Pricing
1.01615
Agar,
extra pure
Expand
View Pricing
1.01614
Agar,
granulated, purified and free from inhibitors, suitable for microbiology
Expand
View Pricing
A9799
Agar,
High gel strength, suitable for plant cell culture
Expand
View Pricing
A4550
Agar,
Type A, suitable for plant cell culture
Expand
View Pricing
05039
Agar,
suitable for microbiology
Expand
View Pricing
A7002
Agar,
ash 2.0-4.5%
Expand
View Pricing
A4800
Agar,
Type M, suitable for plant cell culture
Expand
View Pricing
A8678
Agar,
suitable for plant cell culture
Expand
View Pricing
1012214
Agar,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
12177
Agar,
Suitable for inclusion in culture media.
Expand
View Pricing
01916
Agar,
suitable for microbiology
Expand
View Pricing
05038
Agar,
suitable for microbiology
Expand
View Pricing
56763
Agar,
high purity, low ionic content, low gel strenght, suitable for microbiology
Expand
View Pricing