Skip to Content
Merck
All Photos(1)

Documents

A8986

Sigma-Aldrich

Alexidine dihydrochloride

≥95% (HPLC)

Synonym(s):

1,1′-Hexamethylene-bis(5-[2-ethylhexyl]biguanide)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H56N10 · 2HCl
CAS Number:
Molecular Weight:
581.71
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

DMSO: ≥10 mg/mL

storage temp.

−20°C

SMILES string

Cl.Cl.CCCCC(CC)CNC(=N)NC(=N)NCCCCCCNC(=N)NC(=N)NCC(CC)CCCC

InChI

1S/C26H56N10.2ClH/c1-5-9-15-21(7-3)19-33-25(29)35-23(27)31-17-13-11-12-14-18-32-24(28)36-26(30)34-20-22(8-4)16-10-6-2;;/h21-22H,5-20H2,1-4H3,(H5,27,29,31,33,35)(H5,28,30,32,34,36);2*1H

InChI key

BRJJFBHTDVWTCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Alexidine dihydrochloride has been used:
  • as an antiseptic to study its antimicrobial activity in saliva-derived microcosm biofilms
  • as a protein tyrosine phosphatase localized to the mitochondrion 1 (PTPMT1)-specific inhibitor to study its effects on spare respiratory capacity and viability of CD8+ T cells
  • as a PTPMT1 inhibitor to study its antiviral effect on  human cytomegalovirus (HCMV) replication in HCMV-infected human foreskin fibroblast (HFF) cells

Biochem/physiol Actions

Alexidine dihydrochloride is a bisbiguanide compound. It has been studied in the treatment of head and neck cancer. Alexidine dihydrochloride also exhibits antibiofilm and antifungal activity against several fungal species. It causes mitochondrial apoptosis in mammalian cells due to its anti-cancer activity. Alexidine dihydrochloride is a component of oral disinfectant and contact lens solution.
Alexidine dihydrochloride is a potent and selective PTPMT1 (Protein Tyrosine Phosphatase Localized to the Mitochondrion 1) inhibitor. Alexidine increases insulin secretion by isolated rat pancreatic islets.

Features and Benefits

This compound is featured on the Phosphoprotein Phosphatases (Tyrosine) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophia R Schwarz et al.
Clinical oral investigations, 25(5), 2939-2950 (2020-10-10)
The aims of this study were to investigate the antimicrobial efficacy of antiseptics in saliva-derived microcosm biofilms, and to examine phenotypic adaption of bacteria upon repeated exposure to sub-inhibitory antiseptic concentrations. Saliva-derived biofilms were formed mimicking caries- or gingivitis-associated conditions
J A Chawner et al.
The Journal of applied bacteriology, 66(3), 243-252 (1989-03-01)
A comparative study of the growth inhibitory and bactericidal activities of two related bisbiguanide antiseptics, alexidine and chorhexidine is reported. Whilst overall bactericidal activities and MICs were similar, alexidine was more rapid in its action and it is suggested that
J A Chawner et al.
The Journal of applied bacteriology, 66(3), 253-258 (1989-03-01)
Strains of Providencia stuartii with demonstrated resistance towards chlorhexidine did not show such resistance towards either of the related biguanide antiseptics, alexidine or vantocil. Alexidine promoted a significantly faster alteration in the permeability of Escherichia coli cell membranes towards various
Kenneth W Yip et al.
Molecular cancer therapeutics, 5(9), 2234-2240 (2006-09-21)
Despite advances in surgery, radiation, and chemotherapy, novel therapeutics are needed for head and neck cancer treatment. The objective of this current study was to evaluate alexidine dihydrochloride as a novel compound lead for head and neck cancers. Using a
Zeinab Mamouei et al.
mSphere, 3(5) (2018-11-02)
Invasive fungal infections due to Candida albicans, Aspergillus fumigatus, and Cryptococcus neoformans constitute a substantial threat to hospitalized immunocompromised patients. Further, the presence of drug-recalcitrant biofilms on medical devices and emergence of drug-resistant fungi, such as Candida auris, introduce treatment

Articles

Protein tyrosine phosphatases' catalytic mechanism involves transient phosphorylation.

Protein tyrosine phosphatases' catalytic mechanism involves transient phosphorylation.

Protein tyrosine phosphatases' catalytic mechanism involves transient phosphorylation.

Protein tyrosine phosphatases' catalytic mechanism involves transient phosphorylation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service