Skip to Content
Merck
All Photos(3)

Key Documents

A7883

Sigma-Aldrich

5α-Androst-16-en-3α-ol

Synonym(s):

Androstenol, 16-(5α)Androsten-3α-ol, 3α-Hydroxy-5α-androst-16-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H30O
CAS Number:
Molecular Weight:
274.44
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥98% (TLC)

form

powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

GC/MS: suitable

mp

140-141 °C (lit.)

solubility

ethanol: 19.60-20.40 mg/mL, clear, colorless

shipped in

ambient

storage temp.

room temp

SMILES string

C[C@]12CC[C@H]3[C@@H](CC[C@H]4C[C@H](O)CC[C@]34C)[C@@H]1CC=C2

InChI

1S/C19H30O/c1-18-9-3-4-16(18)15-6-5-13-12-14(20)7-11-19(13,2)17(15)8-10-18/h3,9,13-17,20H,4-8,10-12H2,1-2H3/t13-,14+,15-,16-,17-,18-,19-/m0/s1

InChI key

KRVXMNNRSSQZJP-PHFHYRSDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-α-androst-16-en-3-α-ol (androstenol) is an androgen believed to act as a pheromone. Androstenol has been used in a study to develop a combined gas chromatography/thermal conversion/isotope ratio mass spectrometry (GC/TC/IRMS) method for D/H ratio determination of endogenous urinary steroids.

Biochem/physiol Actions

5alpha-androst-16-en-3alpha-ol (androstenol), is an androgen believed to act as a pheromone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jeremy Vincent et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 61(Pt 1), 156-159 (2006-03-02)
The constitutive androstane receptor (CAR) is a member of the nuclear receptor superfamily. In contrast to classical nuclear receptors, which possess small-molecule ligand-inducible activity, CAR exhibits constitutive transcriptional activity in the apparent absence of ligand. CAR is among the most
Guan Wang et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 870(2), 209-215 (2008-07-01)
A novel method using solid-phase extraction coupled with gas chromatography and flame ionization detector (FID)/electron impact mass spectrometry (EIMS) was developed for the determination of 5alpha-androst-16-en-3alpha-ol (androstenol), a steroidal compound belonging to the group of musk odorous 16-androstenes, in truffle
J J Cowley et al.
The Journal of steroid biochemistry and molecular biology, 39(4B), 647-659 (1991-10-01)
Student volunteers (38 of each sex) were exposed unknowingly overnight to the vapour of pheromonally active substances and compared with controls. The substances were either 5 alpha-16-androsten-3 alpha-ol (androstenol, occurring in human underarm sweat, and known to be pheromonally active
W D Booth
The Journal of endocrinology, 100(2), 195-202 (1984-02-01)
Submaxillary glands of mature Göttingen miniature pigs were examined for the presence of a sexual dimorphism. Gland weights, serous cell hypertrophy and total protein in the glands were much greater in male than female pigs. High concentrations of the pheromonal
D Benton et al.
Biological psychology, 22(2), 141-147 (1986-04-01)
The hypothesis that 5-alpha-androst-16-en-3 alpha-ol increases the sexual arousal of the human female was examined. This substance is produced by the male and has been suggested to be a possible human pheromone. Groups of female subjects were asked to read

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service