Skip to Content
Merck
All Photos(1)

Key Documents

A7650

Sigma-Aldrich

Anti-Goat IgG (whole molecule)–Alkaline Phosphatase antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Rabbit Anti-Goat IgG (whole molecule)–AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

Pricing and availability is not currently available. Contact Technical Service

biological source

rabbit

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:7,000

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

An immunoglobulin has two heavy chains and two light chains connected by disulfide bond.(5) It is a glycoprotein.IgG is a major class of immunoglobulin.(6) Goat IgG has two subclasses- IgG1 and IgG2.(7)
Second antibodies (secondary antibodies) are antibodies raised against other antibodies, typically from another species, called primary antibodies. Primary antibodies are generated with and bind to specific antigens to create antigen antibody complexes which may be detected in a variety of immunochemical and immunohistochemical configurations involving soluble or insoluble signaling molecules. The immunodetection system depends upon the binding of a signal competent secondary antibody to a primary antibody:antigen complex.
The suitability of any secondary antibody to a specific immunodetection application is dependent upon its signal development system configuration. Signal development systems are based upon the chemical conjugation of a signal mediator to the secondary antibody. Major secondary antibody signal systems are color (chromogenic), chemiluminescence or fluorescence based and involve the use of enzymes or high affinity binary systems such as biotin:avidin.

Specificity

Antiserum is determined to be immunospecific for goat IgG by immunoelectrophoresis (IEP), prior to conjugation, versus goat IgG and normal goat serum.

Immunogen

Antiserum is produced in rabbit using purified goat IgG as the immunogen

Application

Anti-Goat IgG (whole molecule)−Alkaline Phosphatase antibody produced in rabbit has been used in western blotting.[1]
Anti-Goat IgG (whole molecule)-Alkaline Phosphatase antibody produced in rabbit has been used in immunoblotting and enzyme-linked immunosorbent assay (ELISA).
Immunocytochemistry was performed on cells fixed in 4% paraformaldhyde for 15 minutes using alkaline phosphatase-conjugated rabbit anti-goat IgG diluted 1:1000 in TBS as the secondary antibody.

Biochem/physiol Actions

IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to the fetus through the placenta that is vital for immune defense of the neonate against infections.
Immunoglobulin G (IgG) mainly helps in immune defense.(5) IgG participates in hypersensitivity type II and type III.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 10 mM glycine, 1% bovine serum albumin, 1 mM MgCl2, 50% glycerol, and 15 mM sodium azide.

Storage and Stability

Store at 2-8°C.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MCMV triggers ROS/NLRP3-associated inflammasome activation in the inner ear of mice and cultured spiral ganglion neurons, contributing to sensorineural hearing loss
Zhuang W, et al.
International Journal of Molecular Medicine, 41(6), 3448-3456 (2018)
Yalan Li et al.
Antioxidants & redox signaling, 38(1-3), 115-136 (2022-06-17)
Aims: Noise damage to auditory hair cells is associated with oxidative stress and mitochondrial dysfunction. This study aimed to investigate the possible effect of sestrin 2 (SESN2), an endogenous antioxidant protein, on noise-induced hearing loss (NIHL) and the underlying mechanisms.
Wei Zhuang et al.
International journal of molecular medicine, 41(6), 3448-3456 (2018-03-08)
Congenital cytomegalovirus (CMV) infection is the most common infectious cause of sensorineural hearing loss in children. While the importance of CMV‑induced SNHL has been described, the mechanisms underlying its pathogenesis and the role of inflammatory responses remain elusive. The present
M Müller et al.
The European respiratory journal, 28(3), 513-522 (2006-06-02)
The increased number of lymphocytes in airways during an asthmatic response is believed to be the result of increased recruitment of these cells. However, it is possible that a decreased apoptotic rate could also contribute to the increased number. The
Placental transfer of IgG subclasses in a Japanese population.
Hashira S, et al.
Pediatrics International : Official Journal of the Japan Pediatric Society, 42(4), 337-342 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service