Skip to Content
Merck
All Photos(1)

Key Documents

A5588

Sigma-Aldrich

Anti-polyHistidine−Alkaline Phosphatase antibody, Mouse monoclonal

clone HIS-1, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-polyHistidine, Monoclonal Anti-polyHistidine−Alkaline Phosphatase antibody produced in mouse, Monoclonal 6 His epitope tag, Monoclonal 6xHis-tag, Monoclonal HHHHHH epitope tag, Monoclonal Hexa His tag, Monoclonal His-tag, Monoclonal His6 tag, Monoclonal Histidine tagged, Monoclonal Poly-His-tag

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.56

biological source

mouse

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

HIS-1, monoclonal

form

buffered aqueous glycerol solution

technique(s)

western blot: 1:2,000 using lysates of Escherichia coli induced to express a 6xHis tagged protein

isotype

IgG2a

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

The Monoclonal Anti-polyHistidine-Alkaline Phosphatase antibody recognizes native and denatured-reduced forms of synthetic polyhistidine or polyhistidine tagged fusion proteins. The product is reactive with fusion protein expressed by prokaryotic pET, pRSET and pTrc expression vectors.

Specificity

The antibody recognizes synthetic polyHistidine, as well as native or denatured, reduced forms of proteins tagged with 6X histidines, expressed in selected vectors.

Immunogen

recombinant polyHistidine tagged fusion protein.

Application

Antibody is suitable for ELISA, immunoblotting and dot blotting assays.

Physical form

Solution in 0.05 M Tris buffer, pH 8.0, containing 1 mM MgCl2, 1% bovine serum albumin, 50% glycerol and 15 mM sodium azide

Legal Information

This Product is covered by patent DE 19507166 and foreign equivalents exclusively licensed to QIAGEN GmbH, Qiagen Strasse 1, D-40724 Hilden, Germany.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of a Short, Disordered N-Terminal Flagellin Segment, a Fully Functional Flagellar Type III Export Signal, to Expression of Secreted Proteins
Dobo,J et al.
Applied and Environmental Microbiology, 6(3), 891-899 (2010)
Rozbeh Jafari et al.
Microbial cell factories, 10, 34-34 (2011-05-18)
Optimization of conditions during recombinant protein production for improved yield is a major goal for protein scientists. Typically this is achieved by changing single crucial factor settings one at a time while other factors are kept fixed through trial-and-error experimentation.
Sarah E Partovi et al.
The Journal of biological chemistry, 293(14), 5236-5246 (2018-02-08)
For nearly 30 years, coenzyme M (CoM) was assumed to be present solely in methanogenic archaea. In the late 1990s, CoM was reported to play a role in bacterial propene metabolism, but no biosynthetic pathway for CoM has yet been
Pooja Bhardwaj et al.
Antimicrobial agents and chemotherapy, 60(4), 2209-2221 (2016-01-27)
Chlorhexidine is a bisbiguanide antiseptic used for infection control. Vancomycin-resistantE. faecium(VREfm) is among the leading causes of hospital-acquired infections. VREfm may be exposed to chlorhexidine at supra- and subinhibitory concentrations as a result of chlorhexidine bathing and chlorhexidine-impregnated central venous
Min Yang et al.
Nature chemical biology, 14(12), 1109-1117 (2018-11-14)
The elucidation and prediction of how changes in a protein result in altered activities and selectivities remain a major challenge in chemistry. Two hurdles have prevented accurate family-wide models: obtaining (i) diverse datasets and (ii) suitable parameter frameworks that encapsulate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service