Skip to Content
Merck
All Photos(1)

Key Documents

A5159

Sigma-Aldrich

Aminopterin

Hybri-Max, 50 ×, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma

Synonym(s):

(S)-2-{4-[(2,4-Diaminopteridin-6-yl)methylamino]benzamido}pentanedioic acid, 4-Amino-PGA, 4-Aminofolic acid, 4-Aminopteroyl-L-glutamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20N8O5
CAS Number:
Molecular Weight:
440.41
Beilstein:
69045
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.75
Pricing and availability is not currently available.

biological source

synthetic

grade

Hybri-Max

sterility

γ-irradiated

product line

BioXtra

form

lyophilized powder

concentration

50 ×

technique(s)

cell culture | hybridoma: suitable

impurities

endotoxin, tested

solubility

10 mL/vial, clear, red (in TC Medium)

ε (extinction coefficient)

24,500 at 282 nm in 0.1 M NaOH at 1 M
25,700 at 261 nm in 0.1 M NaOH at 1 M
8,100 at 373 nm in 0.1 M NaOH at 1 M

shipped in

dry ice

storage temp.

−20°C

SMILES string

Nc1nc(N)c2nc(CNc3ccc(cc3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)cnc2n1

InChI

1S/C19H20N8O5/c20-15-14-16(27-19(21)26-15)23-8-11(24-14)7-22-10-3-1-9(2-4-10)17(30)25-12(18(31)32)5-6-13(28)29/h1-4,8,12,22H,5-7H2,(H,25,30)(H,28,29)(H,31,32)(H4,20,21,23,26,27)/t12-/m0/s1

InChI key

TVZGACDUOSZQKY-LBPRGKRZSA-N

Gene Information

human ... FPGS(2356)
mouse ... Fpgs(14287)

Looking for similar products? Visit Product Comparison Guide

Application

Aminopterin concentrate supplement for hybridoma cell culture applications.

Biochem/physiol Actions

Folic acid antagonist that binds to dihydrofolate reductase and inhibits its activity.
Folic acid antagonist. Aminopterin is actively transported into cells by the folate transporter. In the cell, it is converted to a high molecular weight polyglutamate metabolite by folylpolyglutamate synthase that, in turn, binds to dihydrofolate reductase and inhibits its activity. Aminopterin-polyglutamate is degraded intracellularly by γ-glutamyl hydrolase.

Linkage

More potent, but more toxic, than methotrexate.

Reconstitution

Reconstitute contents of vial with 10 mL sterile cell culture medium. Stock solution is sufficient to prepare 500 mL medium. Final working concentration: 0.4 μM aminopterin.

Legal Information

Hybri-Max is a trademark of Sigma-Aldrich Co. LLC

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Oral - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aleem Gangjee et al.
Journal of medicinal chemistry, 48(16), 5329-5336 (2005-08-05)
We report, for the first time, the biological activities of four-carbon-atom bridged classical antifolates on dihydrofolate reductase (DHFR), thymidylate synthase (TS), and folylpolyglutamate synthetase (FPGS) as well as antitumor activity. Extension of the bridge homologation studies of classical two-carbon bridged
A Rosowsky
Current medicinal chemistry, 6(4), 329-352 (1999-04-02)
Nonpolyglutamatable antifolates are potentially of therapeutic interest for the treatment of tumors that are inherently refractory, or have become resistant, to classical antifolates as a result of decreased expression of the enzyme folylpolyglutamate synthetase. An interesting class of water-soluble nonpolyglutamatable
M Del Campo et al.
Teratology, 60(1), 10-12 (1999-07-21)
Maternal exposures to aminopterin and methotrexate have been associated with a pattern of malformation which includes prenatal-onset growth deficiency, severe lack of ossification of the calvarium, hypoplastic supraorbital ridges, small, low-set ears, micrognathia, and limb abnormalities. We report on a
Steven M Horwitz et al.
Blood, 119(18), 4115-4122 (2012-03-08)
Systemic treatment for cutaneous T-cell lymphoma (CTCL) involves the use of less aggressive, well-tolerated therapies. Pralatrexate is a novel antifolate with high affinity for reduced folate carrier-1. A dose de-escalation strategy identified recommended pralatrexate dosing for patients with CTCL that
Alan Menter et al.
The Journal of pharmacology and experimental therapeutics, 342(3), 696-708 (2012-06-02)
N-[4-[[(2,4-diamino-6-pterdinyl)methyl]amino]benzoyl]-L/D-glutamic acid (L/D-AMT) is an investigational drug in phase 1 clinical development that consists of the L-and D-enantiomers of aminopterin (AMT). L/D-AMT is obtained from a novel process for making the L-enantiomer (L-AMT), a potent oral antiinflammatory agent. The purpose

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service