Skip to Content
Merck
All Photos(1)

Key Documents

A4914

Sigma-Aldrich

Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Rabbit IgG (whole molecule)–HRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:10,000

application(s)

research pathology

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

IgGs are glycoprotein antibodies that modulate several immune responses. Rabbit polyclonal IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary polyclonal anti-rabbit IgGs conjugated to a detectable substrate are useful tools for the immunological analysis of target proteins. Anti-Rabbit IgG (whole molecule)-Peroxidase antibody is specific for all rabbit IgGs and does not bind to human serum proteins in ELISA.

Immunogen

Purified rabbit IgG

Application

Anti-Rabbit IgG (whole molecule)-Peroxidase antibody can be used in sandwich ELISA (diluted 1:3000 in PBS-0.05% Tween 20) and immunoelectrophoresis. The product can also be used for western blotting applications (at 1:2000 dilutions in mouse liver homogenates , or in mouse embryonic fibroblasts ).
Co-immunoprecipation and western blot analysis of C33A cell lysates were performed using HRP conjugated goat anti-rabbit IgG as the secondary antibody.

Other Notes

Antibody adsorbed with human serum proteins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4 containing 1% bovine serum albumin with preservative.

Preparation Note

Prepared using the periodate method described by Wilson, M.B., and Nakane, P.K., in Immunofluorescence and Related Staining Techniques, Elsevier/North Holland Biomedical Press, Amsterdam, p215 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Javeed Jattan et al.
Journal of lipid research, 58(5), 853-865 (2017-02-06)
Since its initial report in 2009, the intestinal enteroid culture system has been a powerful tool used to study stem cell biology and development in the gastrointestinal tract. However, a major question is whether enteroids retain intestinal function and physiology.
Joana Antonela Asensio et al.
Reproductive biology and endocrinology : RB&E, 16(1), 35-35 (2018-04-11)
Allopregnanolone is a neurosteroid synthesized in the central nervous system independently of steroidogenic glands; it influences sexual behavior and anxiety. The aim of this work is to evaluate the indirect effect of a single pharmacological dose of allopregnanolone on important
Lieneke I Bouwman et al.
Journal of immunology (Baltimore, Md. : 1950), 193(9), 4548-4557 (2014-10-01)
The Gram-negative pathogen Campylobacter jejuni is the most common cause of bacterial foodborne disease worldwide. The mechanisms that lead to bacterial invasion of eukaryotic cells and massive intestinal inflammation are still unknown. In this study, we report that C. jejuni
H Sadri et al.
Journal of dairy science, 99(8), 6702-6713 (2016-05-18)
The mammalian target of rapamycin (mTOR) is a major regulator of protein synthesis, whereas the ubiquitin-proteasome system (UPS) is regarded as the main proteolytic pathway in skeletal muscle. The objective of the current study was to investigate the effects of
J Saleh et al.
Journal of lipid research, 39(4), 884-891 (1998-04-29)
The objective of this study was to determine whether Acylation Stimulating Protein (ASP) is generated in vivo by human adipose tissue during the postprandial period. After a fat meal, samples from 12 subjects were obtained (up to 6 h) from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service