Skip to Content
Merck
All Photos(1)

Key Documents

A4727

Sigma-Aldrich

Antisauvagine-30 trifluoroacetate salt

Synonym(s):

Anti-SVG-30, [D-Phe11,His12]-Sauvagine fragment 11-40 trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C161H274N48O46S
Molecular Weight:
3650.26
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

form

powder

Quality Level

storage temp.

−20°C

SMILES string

FC(F)(F)C(=O)O.S(CC[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H](N)Cc2ccccc2)Cc1nc[nH]c1)CC(C)C)CC(C)C)CCCNC(=N)N)CCCCN)C(=O)N[C@@H]([C@H](CC)C)C(=O)N[C@@H](CCC(=O)O)C(=O)N[C@@H]([C@H](CC)C)C(=O)N[C@@H](CCC(=O)O)C(=O)N

InChI

1S/C161H275N49O45S.C2HF3O2/c1-21-84(14)126(130(173)226)207-159(255)129(89(19)211)210-156(252)115(76-125(224)225)206-152(248)111(71-83(12)13)203-150(246)109(69-81(8)9)202-149(245)108(68-80(6)7)200-140(236)98(44-36-65-180-161(176)177)189-154(250)114(75-121(

Gene Information

human ... CRHR2(1395)
mouse ... CRHR2(12922)
rat ... CRHR2(64680)

Amino Acid Sequence

Phe-His-Leu-Leu-Arg-Lys-Met-Ile-Glu-Ile-Glu-Lys-Gln-Glu-Lys-Glu-Lys-Gln-Gln-Ala-Ala-Asn-Asn-Arg-Leu-Leu-Leu-Asp-Thr-Ile-NH2

Biochem/physiol Actions

High-affinity antagonist for Corticotropin-releasing factor (CRF) receptor type 2 (CRF(2)).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nina C Donner et al.
Psychoneuroendocrinology, 63, 178-190 (2015-10-12)
Anxiety and affective disorders are often associated with hypercortisolism and dysfunctional serotonergic systems, including increased expression of TPH2, the gene encoding the rate-limiting enzyme of neuronal serotonin synthesis. We previously reported that chronic glucocorticoid exposure is anxiogenic and increases rat

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service