Skip to Content
Merck
All Photos(1)

Documents

C6499

Sigma-Aldrich

CTB

≥98% (HPLC)

Synonym(s):

CTPB, N-(4-Chloro-3-trifluoromethyl-phenyl)-2-ethoxy-benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13ClF3NO2
CAS Number:
Molecular Weight:
343.73
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

General description

CTB (N-(4-Chloro-3-trifluoromethyl-phenyl)-2-ethoxy-benzamide) is derived from anacardic acid.

Application

CTB has been used to study its apoptotic effects on P53 protein acetylation.

Biochem/physiol Actions

CTPB was a first known small molecular activator of histone acetyltransferase p300. CTB is a simplified analog of CTPB with comparable or higher activity. The compound activates the histone acetyltransferase (HAT) activity of p300/EP300/E1A binding protein.
Small molecule activators of HAT such as CTB (N-(4-Chloro-3-trifluoromethyl-phenyl)-2-ethoxy-benzamide) is useful for altering chromatin acetylation directly. It might serve as a therapeutic agent for treating the complications of cognitive dysfunction including neuropsychiatric, neurodevelopmental and neurodegenerative diseases.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Willian S Korim et al.
Cardiovascular research, 115(6), 1092-1101 (2018-10-26)
Neuroinflammation is a common feature in renovascular, obesity-related, and angiotensin II mediated hypertension. There is evidence that increased release of the pro-inflammatory cytokine tumour necrosis factor-α (TNF-α) contributes to the development of the hypertension, but the underlying neural mechanisms are
Cory Chew et al.
Neurorehabilitation and neural repair, 33(8), 656-667 (2019-07-10)
Background. Motoneuron loss is a severe medical problem that can result in loss of motor control and eventually death. We have previously demonstrated that partial motoneuron loss can result in dendritic atrophy and functional deficits in nearby surviving motoneurons, and
Franklin D Echevarria et al.
Frontiers in neuroscience, 11, 318-318 (2017-06-18)
The pleotropic cytokine interleukin-6 (IL-6) is implicated in retinal ganglion cell (RGC) survival and degeneration, including that associated with glaucoma. IL-6 protects RGCs from pressure-induced apoptosis
The effect of CTB on P53 protein acetylation and consequence apoptosis on MCF-7 and MRC-5 cell lines
Dastjerdi MN, et al.
Advanced Biomedical Research, 2 (2013)
DeMetris D Reed et al.
Meat science, 123, 205-210 (2016-10-19)
Palatability attributes of beef striploin steaks mechanically enhanced with pork fat were evaluated. Striploins were collected from USDA Standard steer carcasses, longitudinally cut into halves (lateral or medial) and assigned randomly to pork fat injection (PFI) or non-injected control (CON).

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service