Skip to Content
Merck
All Photos(1)

Key Documents

A4625

Sigma-Aldrich

N-Acetyl-L-alanine

~99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO3
CAS Number:
Molecular Weight:
131.13
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

N-Acetyl-L-alanine, ~99%

Assay

~99%

form

powder

color

white to off-white

application(s)

cell analysis

storage temp.

2-8°C

SMILES string

C[C@H](NC(C)=O)C(O)=O

InChI

1S/C5H9NO3/c1-3(5(8)9)6-4(2)7/h3H,1-2H3,(H,6,7)(H,8,9)/t3-/m0/s1

InChI key

KTHDTJVBEPMMGL-VKHMYHEASA-N

Application

N-Acetyl-L-alanine has been used to study its effect on cytosolic and mitochondrial reduction-oxidation sensitive green fluorescent protein (roGFP).

Biochem/physiol Actions

N-Acetyl-L-alanine may be used with other L-aminoacylated amino acids as a substrate for the identification, differentiation and characterization of aminoacylase(s)/amidohydrolase(s).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved sensitivity of HSQC spectra of exchanging protons at short interscan delays using a new fast HSQC (FHSQC) detection scheme that avoids water saturation.
S Mori et al.
Journal of magnetic resonance. Series B, 108(1), 94-98 (1995-07-01)
S Mori et al.
Journal of biomolecular NMR, 7(1), 77-82 (1996-01-01)
A technique for separating intramolecular NOE and solvent-proton exchange peaks in exchange spectroscopy is demonstrated. This method utilizes the large differences in relaxation and coupling properties of water and macromolecules to separate the two effects. The spin-echo filter consists of
Bram Boeckx et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 86, 366-374 (2011-11-18)
The conformational landscape of N-acetylalanine has been investigated by a theoretical and matrix-isolation FT-IR study. Optimizations of N-acetylalanine structures has been conducted at successive higher levels of theory HF/3-21G, DFT(B3LYP)/6-31++G** and MP2/6-31++G**. This resulted in three stable conformations. Among these
Linda Chang et al.
Biological psychiatry, 62(12), 1396-1404 (2007-08-19)
Abnormalities in the white matter of the brain may occur in individuals with schizophrenia as well as with normal aging. Therefore, elderly schizophrenic patients may suffer further cognitive decline as they age. This study determined whether elderly schizophrenia participants, especially
Kevin Bray et al.
PloS one, 7(7), e41831-e41831 (2012-08-01)
mTOR inhibitors are used clinically to treat renal cancer but are not curative. Here we show that autophagy is a resistance mechanism of human renal cell carcinoma (RCC) cell lines to mTOR inhibitors. RCC cell lines have high basal autophagy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service