Skip to Content
Merck
All Photos(1)

Key Documents

A4541

Sigma-Aldrich

Anti-Biotin−Peroxidase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Biotin, Anti Biotin Hrp

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

goat

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:2,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Biotin is an essential vitamin required for cells in living organisms and in cultures. The high binding affinity of biotin to egg white avidin or bacteria-derived streptavidin has been used to design immunoassays for detecting antigen-antibody interactions. Goat anti-biotin-peroxidase antibody can be used in many applications where biotin may be introduced as target label.
Specificity and reactivity are determined by Ouchterlony Double Diffusion and immunoelectrophoresis versus the biotin-carrier protein complex. No reactivity is observed versus the carrier protein.

Immunogen

Biotin-KLH conjugate

Application

Biotin-labeled proteins in U937 and BHK cell lysates were analyzed by western blot using HRP-conjugated goat anti-biotin.
Goat anti-biotin-peroxidase antibody can be used for direct ELISA assays at a dilution of 1:2,000.
In some applications, localization of biotinylated probes with avidin produces high background levels. Anti-biotin reagents may be substituted for avidin to decrease non-specific binding.
May be substituted for avidin to decrease non-specific binding.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin with preservative.

Preparation Note

Prepared by the periodate method described by Wilson, M.B., and Nakane, P.K., in "Immunofluorescence and Related Staining Techniques," Elsevier/North Holland Biomedical Press, Amsterdam, p215 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katarzyna Kwiatkowska et al.
Journal of cell science, 116(Pt 3), 537-550 (2003-01-01)
Activation of Fcgamma receptor II (FcgammaRII) induces rearrangement of the actin-based cytoskeleton that serves as a driving force for FcgammaRII-mediated phagocytosis and FcgammaRII capping. To get insight into the signaling events that lead to the actin reorganization we investigated the
Soledad Astrada et al.
Journal of peptide science : an official publication of the European Peptide Society, 22(11-12), 711-722 (2016-12-10)
Because of resistance development by cancer cells against current anticancer drugs, there is a considerable interest in developing novel antitumor agents. We have previously demonstrated that CIGB-552, a novel cell-penetrating synthetic peptide, was effective in reducing tumor size and increasing
Samuel H Henager et al.
Nature methods, 13(11), 925-927 (2016-11-01)
Expressed protein ligation is a valuable method for protein semisynthesis that involves the reaction of recombinant protein C-terminal thioesters with N-terminal cysteine (N-Cys)-containing peptides, but the requirement of a Cys residue at the ligation junction can limit the utility of
Nataliya Gorinski et al.
The Journal of biological chemistry, 295(18), 5970-5983 (2020-03-19)
Barttin is the accessory subunit of the human ClC-K chloride channels, which are expressed in both the kidney and inner ear. Barttin promotes trafficking of the complex it forms with ClC-K to the plasma membrane and is involved in activating
Ee Lin Wong et al.
Nature communications, 10(1), 66-66 (2019-01-10)
Protein-templated fragment ligations have been established as a powerful method for the assembly and detection of optimized protein ligands. Initially developed for reversible ligations, the method has been expanded to irreversible reactions enabling the formation of super-additive fragment combinations. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service