Skip to Content
Merck
All Photos(1)

Key Documents

A3151

Sigma-Aldrich

Avidin–Peroxidase

lyophilized powder

Synonym(s):

Avidin-POD

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

avidin from egg white
enzyme from horseradish

Quality Level

conjugate

peroxidase conjugate

form

lyophilized powder

composition

Protein, ≥70% E1%/280

extent of labeling

0.7-2.0 mol peroxidase per mol avidin

technique(s)

direct ELISA: 1:50,000

solubility

H2O: soluble 1 mg/mL
PBS: soluble

storage temp.

−20°C

Related Categories

General description

Avidin is homotetrameric protein (68kDa) that is obtained from egg whites. This egg protein binds strongly to biotin. Thus, avidin-biotin association has been utilized in immunoassays to detect the localization of antigens in tissues[1]. The use of avidin-biotin immunoassay enhances the sensitivity of the technique and facilitates the detection of antigens in low quantities.

Application

Avidin-Peroxidase has been used for detecting anti-5-bromodeoxyuridine (anti-BrdU) binding[2] and OVA-IgG1[3].
To detect Ga1-deficient IgA1 antibodies in mouse serum, ELISA assays were performed using sera which was incubated with biotinylated HAA, which binds IgA1, in 96-well plates. HRP-conjugated avidin was then incubated with the sera for 40 minutes at 37 degrees.

Physical form

Lyophilized powder containing citrate buffer.

Preparation Note

Labeled with Type VI peroxidase. Coupled by a modification of the method of Sullivan, M.J., et al., FEBS Lett., 95, 311 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Avia Merenlender-Wagner et al.
Autophagy, 10(12), 2324-2332 (2014-12-09)
Autophagy plays a key role in the pathophysiology of schizophrenia as manifested by a 40% decrease in BECN1/Beclin 1 mRNA in postmortem hippocampal tissues relative to controls. This decrease was coupled with the deregulation of the essential ADNP (activity-dependent neuroprotector
D G Alleva et al.
Diabetes, 49(7), 1106-1115 (2000-07-26)
Cytokines derived from macrophages (Mø) play a critical role in the development of type 1 diabetes in the nonobese diabetic (NOD) mouse. Based on earlier findings from lupus-prone strains of inherent cytokine defects in Mø , NOD Mø were evaluated
Anton B Iliuk et al.
Current protocols in chemical biology, 7(1), 17-25 (2015-03-03)
This article describes methods for a new, non-antibody phosphorylation detection reagent, termed pIMAGO (phospho-imaging). This novel reagent takes advantage not only of the unique properties of the soluble nanoparticles, but also of the multiple functionalities of the molecule, allowing for
Luigia Pace et al.
Journal of immunology (Baltimore, Md. : 1950), 184(11), 5969-5979 (2010-04-30)
Type I IFNs are central to a vast array of immunological functions. Their early induction in innate immune responses provides one of the most important priming mechanisms for the subsequent establishment of adaptive immunity. The outcome is either promotion or
P Birner et al.
The American journal of pathology, 158(6), 1991-1996 (2001-06-08)
Monoclonal antibody MIB-1 is a reliable tool for determining proliferating cells in human tissues, but does not react with the homologous mouse antigen and is therefore useless in experimental pathology using mice as model systems. Standard method for assessment of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service