Skip to Content
Merck
All Photos(1)

Documents

A4262

Sigma-Aldrich

Actinomycin D

from Streptomyces sp., ≥95% (HPLC)

Synonym(s):

Actinomycin C1, Actinomycin IV, Dactinomycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H86N12O16
CAS Number:
Molecular Weight:
1255.42
Beilstein:
605235
EC Number:
MDL number:
UNSPSC Code:
51282001
PubChem Substance ID:
NACRES:
NA.76

biological source

Streptomyces sp.

Quality Level

Assay

≥95% (HPLC)

form

powder

color

red, powder

mp

251-253

solubility

ethanol, DMSO: soluble

antibiotic activity spectrum

neoplastics

Mode of action

DNA synthesis | interferes

storage temp.

2-8°C

SMILES string

CC(C)[C@H]1NC(=O)[C@@H](NC(=O)c2ccc(C)c3OC4=C(C)C(=O)C(N)=C(C(=O)N[C@H]5[C@@H](C)OC(=O)[C@H](C(C)C)N(C)C(=O)CN(C)C(=O)[C@@H]6CCCN6C(=O)[C@H](NC5=O)C(C)C)C4=Nc23)[C@@H](C)OC(=O)[C@H](C(C)C)N(C)C(=O)CN(C)C(=O)[C@@H]7CCCN7C1=O

InChI

1S/C62H86N12O16/c1-27(2)42-59(84)73-23-17-19-36(73)57(82)69(13)25-38(75)71(15)48(29(5)6)61(86)88-33(11)44(55(80)65-42)67-53(78)35-22-21-31(9)51-46(35)64-47-40(41(63)50(77)32(10)52(47)90-51)54(79)68-45-34(12)89-62(87)49(30(7)8)72(16)39(76)26-70(14)58(83)37-20-18-24-74(37)60(85)43(28(3)4)66-56(45)81/h21-22,27-30,33-34,36-37,42-45,48-49H,17-20,23-26,63H2,1-16H3,(H,65,80)(H,66,81)(H,67,78)(H,68,79)/t33-,34-,36+,37+,42-,43-,44+,45+,48+,49+/m1/s1

InChI key

RJURFGZVJUQBHK-IIXSONLDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: peptide

Application

Actinomycin D is an antineoplastic antibiotic that inhibits cell proliferation with wide-ranging applications ranging from a selection agent in cell culture, use in studies of suppressing HIV-replication and programmed cell death of PC12 cells. Recommended for us in cell culture applications at 1μg/mL.

Biochem/physiol Actions

Actinomycin D inhibits the proliferation of cells by forming a stable complex with double-stranded DNA, inhibiting DNA-primed RNA synthesis and causing single-stranded breaks in DNA. It has been shown to be an inhibitor of the minus-strand transfer step in reverse transcriptase.

Caution

This product is hygroscopic and light-sensitive. If stored protected from light and moisture at 2-8°C, it remains unchanged, by HPLC testing, for at least 15 months. Unused dilute solutions of Actinomycin D are extremely sensitive to light and tend to adsorb to plastic and glass and should be discarded. However, frozen aliquots of concentrated stock solution are stable for at least a month.

Preparation Note

Actinomycin D is sold as red, shiny crystals and is soluble in acetonitrilie or acetone at 10 mg/mL and in DMSO at a minimum of 1 mg/mL. Some references also show slight solubility in water of 0.5 mg/mL.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Carc. 1B - Repr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio Rescifina et al.
Journal of medicinal chemistry, 49(2), 709-715 (2006-01-20)
Isoxazolidinyl polycyclic aromatic hydrocarbons (isoxazolidinyl-PAHs) have been synthesized in good yields by 1,3-dipolar cycloaddition methodology promoted by microwave irradiation. The structures of the obtained cycloadducts have been determined by NOE experiments and supported by computational studies at the AM1 level.
Leilei Zhou et al.
Genome biology, 20(1), 156-156 (2019-08-08)
Methylation of nucleotides, notably in the forms of 5-methylcytosine (5mC) in DNA and N6-methyladenosine (m6A) in mRNA, carries important information for gene regulation. 5mC has been elucidated to participate in the regulation of fruit ripening, whereas the function of m6A
Arkady Khoutorsky et al.
Neuron, 78(2), 298-311 (2013-04-30)
Control of protein synthesis is critical for synaptic plasticity and memory formation. However, the molecular mechanisms linking neuronal activity to activation of mRNA translation are not fully understood. Here, we report that the translational repressor poly(A)-binding protein (PABP)-interacting protein 2A
Rolf Schreckenberg et al.
Antioxidants & redox signaling, 23(15), 1220-1232 (2015-05-16)
Research into right ventricular (RV) physiology and identification of pathomechanisms underlying RV failure have been neglected for many years, because function of the RV is often considered less important for overall hemodynamics and maintenance of blood circulation. In view of
H Sun et al.
British journal of cancer, 111(2), 281-291 (2014-06-20)
We investigated the biologic and pharmacologic activities of a chromosome region maintenance 1 (CRM1) inhibitor against human non-small cell lung cancer (NSCLC) cells both in vitro and in vivo. The in vitro and in vivo effects of a novel CRM1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service