Skip to Content
Merck
All Photos(5)

Key Documents

A0884

Sigma-Aldrich

L-Asparagine

L-Asparagine

≥98% (HPLC)

Synonym(s):

(S)-2-Aminosuccinic acid 4-amide, L-Aspartic acid 4-amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCOCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
132.12
Beilstein:
1723527
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Product Name

L-Asparagine, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white

mp

235 °C (dec.) (lit.)

application(s)

detection

SMILES string

N[C@@H](CC(N)=O)C(O)=O

InChI

1S/C4H8N2O3/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H2,6,7)(H,8,9)/t2-/m0/s1

InChI key

DCXYFEDJOCDNAF-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-asparagine has been used:
  • to identify and quantify free amino acids released upon oxidation of proteins and peptides by hydroxyl radicals[1]
  • to study the effects of amino acids in promoting food consumption in Drosophila melanogaster[2]
  • to study non-enzymatic gluconeogenesis[3]

Biochem/physiol Actions

L-asparagine is an uncharged derivative of aspartate. It possesses a polar side chain and is a non-essential amino acid.[4][5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Release of free amino acids upon oxidation of peptides and proteins by hydroxyl radicals
Analytical and Bioanalytical Chemistry (2017)
Volker F. Wendisch
Amino Acid Biosynthesis ? Pathways, Regulation and Metabolic Engineering (2007)
Nonenzymatic gluconeogenesis-like formation of
fructose 1,6-bisphosphate in ice
Christoph B. Messner
Proceedings of the National Academy of Sciences of the USA, 114 null
Stryer L
Biochemistry (2002)
An internal sensor detects dietary amino acids and promotes food consumption in Drosophila
Zhe Yang
Nature (2017)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service