Skip to Content
Merck
All Photos(2)

Key Documents

A2918

Sigma-Aldrich

Azure A chloride

Dye content, ≥75%, certified by the Biological Stain Commission, powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14ClN3S
CAS Number:
Molecular Weight:
291.80
Colour Index Number:
52005
Beilstein:
3922287
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Azure A chloride, certified by the Biological Stain Commission

grade

certified by the Biological Stain Commission

Quality Level

form

powder

composition

Dye content, ≥75%

color

dark brown
green to very dark green

mp

290 °C (dec.) (lit.)

solubility

water: 1 mg/mL

suitability

suitable for (Certified for use in Lillie′s modified Nocht′s method for paraffin sections and the NCCLS method for blood smears.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

NC1=CC(SC(C2=N3)=CC(C=C2)=[N+](C)C)=C3C=C1.[Cl-]

InChI

1S/C14H13N3S.ClH/c1-17(2)10-4-6-12-14(8-10)18-13-7-9(15)3-5-11(13)16-12;/h3-8,15H,1-2H3;1H

InChI key

NALREUIWICQLPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Suitability

Certified for use in Lillie′s modified Nocht′s method for paraffin sections and the NCCLS method for blood smears.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scan-and-treat methodology using Azure A fast stain as a cost-effective cervical cancer screening alternative to visual inspection with acetic acid.
Martial Guillaud et al.
Gynecologic oncology, 107(1 Suppl 1), S256-S259 (2007-09-11)
P Chieco et al.
The Histochemical journal, 25(8), 569-577 (1993-08-01)
Mitotic index is a clinically important parameter in cancer pathology. We developed a staining method using Toluidine Blue to detect efficiently and rapidly mitotic figures in sections of formalin-fixed paraffin-embedded human and rat tissues. Sections were stained at acid pH
F Battaglini et al.
Bioconjugate chemistry, 5(5), 430-435 (1994-09-01)
The bimolecular rate constants for the reactions of five organic two-electron redox mediators with reduced glucose oxidase (GOx) were determined by measuring voltammetric electrocatalytic currents at glassy carbon electrodes in the presence of excess glucose under anaerobic conditions. The mediators
Shuhua Bai et al.
Journal of pharmaceutical sciences, 96(8), 2090-2106 (2007-02-09)
This study was designed to test the hypothesis that positively charged dendrimers form a complex with enoxaparin, a low-molecular weight heparin (LMWH), and that the resulting drug-dendrimer complex is effective in preventing deep vein thrombosis after pulmonary administration. Fourier Transform
J F Liang et al.
International journal of pharmaceutics, 202(1-2), 11-20 (2000-08-01)
A novel heparin/protamine-based approach for delivery of enzyme drugs without associated toxic effects has been proposed. This approach would allow an enzyme drug to be administered in an inactive (i.e. pro-drug) form and then released at the target site in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service