Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Azure A chloride
Empirical Formula (Hill Notation):
C
14
H
14
ClN
3
S
CAS Number:
531-53-3
Molecular Weight:
291.80
EC Number:
208-510-7
Products
Product Category
stains (3)
Brand
Sigma-Aldrich (3)
Color
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Application
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
A6270
Azure A chloride,
Dye content ≥70 %, Powder or crystals
Expand
View Pricing
861049
Azure A chloride,
certified by the Biological Stain Commission, Dye content 70 %
Expand
View Pricing
A2918
Azure A chloride,
Dye content, ≥75%, certified by the Biological Stain Commission, powder
Expand
View Pricing