Skip to Content
Merck
All Photos(3)

Key Documents

A2408

Sigma-Aldrich

S-(5′-Adenosyl)-L-methionine p-toluenesulfonate salt

≥80% (HPLC), ≥80% (spectrophotometric assay)

Synonym(s):

AdoMet, SAM p-toluenesulfonate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23N6O5S · C7H7O3S · xC7H8O3S
CAS Number:
Molecular Weight:
570.64 (free base basis)
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

biological source

synthetic (Organic)

Quality Level

Assay

≥80% (HPLC)
≥80% (spectrophotometric assay)

form

powder

composition

p-Toluenesulfonate content, 1.6-4.4 mol/mol

color

white to off-white

solubility

H2O: 100 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(O[H])[C@](N([H])[H])([H])C([H])([H])C([H])([H])[S@+](C([H])([H])[H])C([H])([H])[C@@]([C@](O[H])([H])[C@@]1([H])O[H])([H])O[C@]1([H])N2C3=C(N=C2[H])C(N([H])[H])=NC([H])=N3.O=S(C4=C([H])C([H])=C(C([H])([H])[H])C([H])=C4[H])([O-])=O

InChI

1S/C15H22N6O5S.C7H8O3S/c1-27(3-2-7(16)15(24)25)4-8-10(22)11(23)14(26-8)21-6-20-9-12(17)18-5-19-13(9)21;1-6-2-4-7(5-3-6)11(8,9)10/h5-8,10-11,14,22-23H,2-4,16H2,1H3,(H2-,17,18,19,24,25);2-5H,1H3,(H,8,9,10)/t7-,8+,10+,11+,14+,27?;/m0./s1

InChI key

VHPOFDUCFKOUHV-XKGORWRGSA-N

General description

S-(5′-Adenosyl)-L-methionine p-toluenesulfonate salt is used for the treatment of depression, liver cirrhosis, cholestasis and degenerative joint disease.

Application

S-(5′-Adenosyl)-L-methionine p-toluenesulfonate salt has been used for cell proliferation assay. It has also been used for methylation assay.

Biochem/physiol Actions

S-(5′-Adenosyl)-L-methionine (SAM) is anti-apoptotic in normal hepatocytes and normal colon epithelial cells but pro-apoptotic in liver human hepatocellular carcinoma (HCC), HepG2 cells and colon cancer cells.
Methyl donor; cofactor for enzyme-catalyzed methylations, including catechol O-methyltransferase (COMT) and DNA methyltransferases (DNMT).
Methyl donor; cofactor for enzyme-catalyzed methylations, including catechol O-methyltransferase (COMT) and DNA methyltransferases (DNMT). Although present in all cells, it is concentrated in liver where 85% of all methylation reactions occur. It is also involved in regulating liver function, growth, and response to injury.

Caution

This material is unstable at room temperature.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Negative regulation of interferon-induced transmembrane protein 3 by SET7-mediated lysine monomethylation
Shan Z, et al.
The Journal of biological chemistry, 288(49), 35093-35103 (2013)
Yang-Liu Xia et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1157, 122333-122333 (2020-09-01)
Catechol-O-methyltransferase (COMT) is a major drug metabolizing enzyme in humans. COMT expression is directedly associated with various mental diseases and cancers due to its essential role in catalyzing metabolic inactivation of endogenous catecholamines and catechol estrogens. However, a practical method
Allison B Norvil et al.
Nucleic acids research, 48(7), 3761-3775 (2020-03-04)
We have previously shown that the highly prevalent acute myeloid leukemia (AML) mutation, Arg882His, in DNMT3A disrupts its cooperative mechanism and leads to reduced enzymatic activity, thus explaining the genomic hypomethylation in AML cells. However, the underlying cause of the
Le Thanh Mai Pham et al.
Enzyme and microbial technology, 82, 66-73 (2015-12-18)
Using bioinformatic homology search tools, this study utilized sequence phylogeny, gene organization and conserved motifs to identify members of the family of O-methyltransferases from lignin-degrading fungus Phanerochaete chrysosporium. The heterologous expression and characterization of O-methyltransferases from P. chrysosporium were studied.
Lupus autoimmunity altered by cellular methylation metabolism
Yang ML, et al.
Autoimmunity, 46(1), 21-31 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service