Skip to Content
Merck
All Photos(1)

Key Documents

A2080

Sigma-Aldrich

Monoclonal Anti-Alkaline Phosphatase, Human Placental−Agarose antibody produced in mouse

clone 8B6, purified immunoglobulin, PBS suspension

Synonym(s):

Monoclonal Anti-Alkaline Phosphatase, Human Placental, Anti-Phosphatase, Alkaline, Human Placental, PLAP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

agarose conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8B6, monoclonal

form

PBS suspension

species reactivity

human

extent of labeling

2 mg antibody per mL bed volume

technique(s)

immunoprecipitation (IP): suitable

isotype

IgG2a

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ALPP(250)

Looking for similar products? Visit Product Comparison Guide

General description

In SDS gels, the product reacts with both Regan and Nagao isozymes of human placental alkaline phosphatase (hPLAP, 130 kDa, 67/130 kDa). By RIA, the antibody binds to hPLAP with an affinity constant of 5 × 109 LM-1. It does not react with PLAP-like enzymes.
Monoclonal Anti-Human Placental Alkaline Phosphatase (mouse IgG2a isotype) is derived from the 8B6 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from immunized BALB/c mice. Alkaline Phosphatase (AP) is a broad and general term associated with non-specific phosphomonoesterases, with optimal activity at alkaline pH. Human placental alkaline phosphatase (EC 3.1.3.1; hPLAP), an isoenzyme of the AP group of enzymes, is ordinarily synthesized in the placental syncytiotrophoblast, becoming detectable in maternal circulation after the twelfth week of pregnancy. Small amounts of hPLAP are also found in the endocervix, fallopian tubes and lung. Very small amounts of heat-stable AP resembling hPLAP (hPLAP-like AP) are expressed in the testis, thymus and in rare colon epithelial cells.

Immunogen

Human epidermoid carcinoma cell line expressing placental alkaline phosphatase.

Application

Anti-Alkaline phosphatase human placental antibody coupled to agarose was packed in a column and used to purify alkaline-phosphatase conjugated FGF receptor out of conditioned media from COS-7 cells. Column was washed with 1M NaCl.
Cultured pancreatic islet cells were fixed with cold 4% paraformaldhyde at room temperature for 10 minutes and incubated with mouse monoclonal anti-Alkaline Phosphatase, Human Placental antibody at 1:500. Mouse monoclonal anti-Alkaline Phosphatase, Human Placental antibody was used to detect a unique molecular probe that consisted of the FGFR extracellular domain expressed as a chimera with human PLAP.
This immunoadsorbant may be used to purify fusion proteins produced using vectors expressing the hPLAP gene. Suitable for affinity purification and immunoprecipitation procedures of human placental alkaline phosphatase either natural or recombinant.It is alsu used in western blotting.

Biochem/physiol Actions

Alkaline phosphatases are non-specific phosphomonoesterases that function optimally at alkaline pH. Placental alkaline phosphatase differs from the alkaline phosphatases of bone, liver and kidney, in the hydrolysis rate of various phosphate-based substrates, and also in its degree of inhibition by L-phenylalanine and L-homoarginine or levamisole. Human Placental Alkaline Phosphatase (hPLAP) has been studied as a potential tumor marker.
The human placental alkaline phosphatase (hPLAP and hPLAP-like AP) is present in the sera and tumor tissue of patients suffering from various types of cancer. Despite the continued interest in hPLAP as a potential tumor marker, it has not been widely used in the routine clinical laboratory, mainly because of the low overall specificity attained by methodologies such as heat-inactivation and sensitivity to L-phenylalanine.

Physical form

Suspension in 0.1 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Preparation Note

Prepared using purified antibody coupled to cyanogen bromide-activated agarose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extremely high maternal alkaline phosphatase serum concentration with syncytiotrophoblastic origin
Boronkai A, et al.
Journal of Clinical Pathology, 58(1), 72-76 (2005)
Songshan Ye et al.
Molecular medicine reports, 13(6), 4593-4598 (2016-04-16)
Paeoniflorin, the major component of Paeonia lactiflora pall, has previously been reported to prevent thrombosis. Plasminogen activator urokinase (uPA) is a serine protease that markedly facilitates normal thrombosis resolution. Paeoniflorin and uPA have been linked to the mitogen‑activated protein kinase (MAPK)
Alkaline phosphatases
Millan JL
Purinergic Signaling, 2(2), 335-335 (2006)
Xinghua Hou et al.
The Journal of biological chemistry, 287(1), 474-483 (2011-11-15)
NOTCH signaling induced by Delta1 (DLL1) and Jagged1 (JAG1) NOTCH ligands is modulated by the β3N-acetylglucosaminyl transferase Fringe. LFNG (Lunatic Fringe) and MFNG (Manic Fringe) transfer N-acetylglucosamine (GlcNAc) to O-fucose attached to EGF-like repeats of NOTCH receptors. In co-culture NOTCH
Specific immunoassays for placental alkaline phosphatase as a tumor marker
Stinghen S, et al.
BioMed Research International, 2006 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service