Skip to Content
Merck
All Photos(1)

Key Documents

P0762

Sigma-Aldrich

Phosphatase, Alkaline−Agarose from calf intestine

ammonium sulfate suspension

Synonym(s):

Alkaline phosphatase-Agarose

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
eCl@ss:
42010105
NACRES:
NA.54

biological source

bovine (calf) intestine

form

ammonium sulfate suspension

specific activity

250-1000 units/mL packed gel

matrix

beaded agarose

storage temp.

2-8°C

General description

Alkaline phosphatase from calf intestine is a glycoprotein with a molecular weight of 140,000.

Application

Alkaline phosphatase from calf intestine, CIP, removes phosphate groups from the 5′ ends of DNA which aids in cloning techniques to keep DNA from self-ligation. It can be inhibited by homoarginine.
Calf intestinal alkaline phosphatase (CIAP) has been used in a study with the spectrophotometric method to assess the inhibition effect of dansyl-L-phenylalanine. It has also been shown that (CIAP) is able to detoxify the lipopolysaccharide (LPS)-mediated inflammatory response, which could be important in the treatment of LPS-mediated diseases.

Biochem/physiol Actions

Sigma insoluble enzymes are produced by reacting the conventional "soluble" enzyme with an inert base that results in an insoluble compound retaining the activity of the original enzyme. Applications are almost unlimited. By selecting the "inert" base carefully, a highly active "resin" or "gel" can be produced with which the enzyme reaction can be quickly catalyzed by momentary contact with the substrate in a suitable medium. This can be carried out either in a batch slurry or even through a small column. In either case, the insoluble enzyme does not remain as a contaminant of the reaction mix. It can be readily filtered or centrifuged out of the batch slurry, or remain behind in the column. Many unstable enzymes seem to be much more stable in the insoluble form.

Packaging

Package size based on DEA units

Unit Definition

One unit will hydrolyze 1.0 mmole of p-nitrophenyl phosphate per min at pH9.8 at 37 °C.

Physical form

Suspension in 2.0 M (NH4)2SO4, 1 mM MgCl2, 0.1 mM ZnCl2, pH 7

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Bakker et al.
Journal of virology, 70(8), 5511-5518 (1996-08-01)
The Rex protein is an essential regulator of RNA expression in human T-cell leukemia virus types 1 and 2 (HTLV-1 and HTLV-2) that promotes the accumulation of full-length and partially spliced viral transcripts in the cytoplasm. Rex-mediated regulation correlates with
Rebika Shrestha et al.
iScience, 25(1), 103608-103608 (2022-02-03)
Understanding the spatiotemporal distribution and dynamics of RAS on the plasma membrane (PM) is the key for elucidating the molecular mechanisms of the RAS signaling pathway. Single particle tracking (SPT) experiments show that in cells, KRAS diffuses in at least
Li-Na Li et al.
Guang pu xue yu guang pu fen xi = Guang pu, 29(10), 2820-2823 (2009-12-30)
To evaluate the inhibition effect of dansyl-L-phenylalanine on calf intestinal alkaline phosphatase (CIAP), UV-Vis spectrophotometric method was employed. It was found that dansyl-L-phenylalanine can selectively inhibit CIAP. The kinetic inhibition processes of dansyl-L-phenylalanine and L-phenylalanine were comparatively studied. The authors'
J E Coleman
Annual review of biophysics and biomolecular structure, 21, 441-483 (1992-01-01)
Alkaline phosphatase was the first zinc enzyme to be discovered in which three closely spaced metal ions (two Zn ions and one Mg ion) are present at the active center. Zn ions at all three sites also produce a maximally
M Kellerer et al.
Diabetologia, 41(7), 833-838 (1998-08-01)
Protein kinase C (PKC) isoforms are potentially important as modulators of the insulin signalling chain and could be involved in the pathogenesis of cellular insulin resistance. We have previously shown that phorbol ester stimulated PKC beta1 and beta2 as well

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service