Skip to Content
Merck
All Photos(1)

Documents

A1164

Sigma-Aldrich

Azaserine

Hybri-Max, γ-irradiated, 50x, lyophilized powder, BioXtra, suitable for hybridoma

Synonym(s):

O-Diazoacetyl-L-serine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7N3O4
CAS Number:
Molecular Weight:
173.13
Beilstein:
1726602
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

grade

Hybri-Max

Quality Level

sterility

γ-irradiated

product line

BioXtra

form

lyophilized powder

technique(s)

cell culture | hybridoma: suitable

impurities

endotoxin, tested

antibiotic activity spectrum

fungi

Mode of action

enzyme | inhibits

storage temp.

−20°C

SMILES string

N[C@@H](COC(=O)C=[N+]=[N-])C(O)=O

InChI

1S/C5H7N3O4/c6-3(5(10)11)2-12-4(9)1-8-7/h1,3H,2,6H2,(H,10,11)/t3-/m0/s1

InChI key

MZZGOOYMKKIOOX-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: amino acid derivatives

Application

For use in hybridoma cell culture applications as a myeloma selection agent. Used to study inhibition of formylglycinamide ribonucleotide amidotransferase and PRPP amidotransferase.

Biochem/physiol Actions

Azaserine is an antibiotic, generated by Streptomyces fragilis. It is considered as a potential mutation inducing agent. It is known to promote pancreatic cancer and induces adenoma and carcinoma in vivo and in vitro. Azaserine also stimulates kidney and liver tumors in rats.

Reconstitution

Reconstitute contents of vial with 10 mL sterile cell culture medium. Stock solution is sufficient to prepare 500 mL medium. Final working concentration: 5.7 μM azaserine.

Legal Information

Hybri-Max is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antineoplastic and Immunosuppressive Agents, Part 2, 494-494 (2013)
Kei Wada et al.
Journal of molecular biology, 380(2), 361-372 (2008-06-17)
gamma-Glutamyltranspeptidase (GGT) catalyzes the cleavage of such gamma-glutamyl compounds as glutathione, and the transfer of their gamma-glutamyl group to water or to other amino acids and peptides. GGT is involved in a number of biological phenomena such as drug resistance
Céline Masclaux-Daubresse et al.
Plant physiology, 140(2), 444-456 (2006-01-13)
Glutamate (Glu) metabolism and amino acid translocation were investigated in the young and old leaves of tobacco (Nicotiana tabacum L. cv Xanthi) using [15N]ammonium and [2-15N]Glu tracers. Regardless of leaf age, [15N]ammonium assimilation occurred via glutamine synthetase (GS; EC 6.1.1.3)
The Pathobiology of Neoplasia, 154-154 (2012)
Tusty-Jiuan Hsieh et al.
The Journal of endocrinology, 183(3), 535-550 (2004-12-14)
We reported previously that insulin inhibits the stimulatory effect of high glucose on the expression of angiotensinogen (ANG) gene in both rat immortalized renal proximal tubular cells (IRPTCs) and non-diabetic rat renal proximal tubular cells (RPTCs), but has no effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service