Skip to Content
Merck
All Photos(1)

Key Documents

A114

Sigma-Aldrich

(+)-N-Allylnormetazocine hydrochloride

≥98% (HPLC)

Synonym(s):

(+)-NANM hydrochloride, SKF-10047, SKF10047, [2S-(2α, 6α,11R*)]-1,2,3,4,5,6-Hexahydro-6,11-dimethyl-3-(2-propenyl)-2,6-methano-3-benzazocin-8-ol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23NO · HCl
CAS Number:
Molecular Weight:
293.83
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

optical activity

[α]21/D +90.6°, c = 1 in ethanol(lit.)

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

storage condition

desiccated

color

white

solubility

H2O: >10 mg/mL

storage temp.

room temp

SMILES string

Cl[H].C[C@@H]1[C@@H]2Cc3ccc(O)cc3[C@@]1(C)CCN2CC=C

InChI

1S/C17H23NO.ClH/c1-4-8-18-9-7-17(3)12(2)16(18)10-13-5-6-14(19)11-15(13)17;/h4-6,11-12,16,19H,1,7-10H2,2-3H3;1H/t12-,16+,17+;/m1./s1

InChI key

ZTGMHFIGNYXMJV-XSCGHNKWSA-N

Gene Information

human ... OPRS1(10280)

Biochem/physiol Actions

Selective σ1 receptor agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R A Wilke et al.
The Journal of biological chemistry, 274(26), 18387-18392 (1999-06-22)
Recent work has indicated that sigma receptor ligands can modulate potassium channels. However, the only sigma receptor characterized at the molecular level has a novel structure unlike any other receptor known to modulate ion channels. This 26-kDa protein has a
Parham Gharagozlou et al.
BMC pharmacology, 3, 1-1 (2003-01-07)
The aim of the present study was to describe the activity of a set of opioid drugs, including partial agonists, in a cell system expressing only mu opioid receptors. Receptor activation was assessed by measuring the inhibition of forskolin-stimulated cyclic
Parham Gharagozlou et al.
BMC pharmacology, 6, 3-3 (2006-01-26)
The aim of the present study was to describe the activity of a set of opioid drugs, including partial agonists, in a human embryonic kidney cell system stably expressing only the mouse kappa-opioid receptors. Receptor activation was assessed by measuring
Mohamed S Amer et al.
British journal of pharmacology, 168(6), 1445-1455 (2012-11-06)
The Sigma-1 receptor (Sig1R) impacts on calcium ion signalling and has a plethora of ligands. This study investigated Sig1R and its ligands in relation to endogenous calcium events of endothelial cells and transient receptor potential (TRP) channels. Intracellular calcium and
C Bucolo et al.
The Journal of pharmacology and experimental therapeutics, 289(3), 1362-1369 (1999-05-21)
In this study, we examined the presence of sigma1 and sigma2 sites in the rabbit iris-ciliary body by receptor binding and investigated their effects on intraocular pressure (IOP) in albino rabbits. The iris-ciliary body has binding sites for the sigma1-site

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service