Skip to Content
Merck
All Photos(1)

Key Documents

95904

Sigma-Aldrich

Poly(ethylene glycol)

BioUltra, 4,000

Synonym(s):

Polyethylene glycol, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.26

product line

BioUltra

form

powder

mol wt

Mr 3500-4500

impurities

insoluble matter:, passes filter test
≤0.001% peroxides (as H2O2):

pH

5.5-7.0 (25 °C, 50 mg/mL in H2O)

mp

58-61 °C

solubility

H2O: 50 mg/mL at 25 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤200 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤200 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

50 mg/mL in H2O

UV absorption

λ: 260 nm Amax: ≤0.10
λ: 280 nm Amax: ≤0.04

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (PEG) is a polymer ethylene glycol units and is hydrophilic in nature. PEG is structurally flexible and has high hydration capacity. PEG is biocompatible and is devoid of steric hindrances in its structure. PEG is a Food and Drug Administration (FDA) approved drug conjugates. It is nonimmunogenic and safe. PEG conjugation improves solubility and dissolution of diabetes drug, Gliclazide. PEG is widely used as a reagent in biochemistry and molecular biology experiments. It is used in carbon nanotube functionalization.

Application

Poly(ethylene glycol) has been used:
  • as a component of wheat protoplast isolation and transformation buffer
  • as an additive in suspension cell culture
  • as a component of yeast transformation buffer
  • as a component of crystallization buffer in cysteine protease ATG4B crystallization

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

281.5 °F - closed cup

Flash Point(C)

138.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterisation of gliclazide-PEG 8000 solid dispersions
Biswal S, et al.
Tropical Journal of Pharmaceutical Research, 8(5), 260-260 (2009)
Adaptation of the Se301 insect cell line to suspension culture. Effect of turbulence on growth and on production of nucleopolyhedrovius (SeMNPV)
Beas-Catena A, et al.
Cytotechnology, 63(6), 543-552 (2011)
ATG4B contains a C-terminal LIR motif important for binding and efficient cleavage of mammalian orthologs of yeast Atg8
Rasmussen MS, et al.
Autophagy, 13(5), 834-834 (2017)
Genome editing of bread wheat using biolistic delivery of CRISPR/Cas9 in vitro transcripts or ribonucleoproteins
Liang Z, et al.
Nature Protocols, 13(3), 413-413 (2018)
Directed molecular evolution of DREADDs: a generic approach to creating next-generation RASSLs
Dong S, et al.
Nature Protocols, 5(3), 561-561 (2010)

Related Content

Polyethylene glycol (PEG), also sometimes referred to as polyethylene oxide (PEO), is a condensation polymer of ethylene oxide and water that has several chemical properties that make it useful for biological, chemical and pharmaceutical applications.

Polyethylene glycol (PEG), also sometimes referred to as polyethylene oxide (PEO), is a condensation polymer of ethylene oxide and water that has several chemical properties that make it useful for biological, chemical and pharmaceutical applications.

Polyethylene glycol (PEG), also sometimes referred to as polyethylene oxide (PEO), is a condensation polymer of ethylene oxide and water that has several chemical properties that make it useful for biological, chemical and pharmaceutical applications.

Polyethylene glycol (PEG), also sometimes referred to as polyethylene oxide (PEO), is a condensation polymer of ethylene oxide and water that has several chemical properties that make it useful for biological, chemical and pharmaceutical applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service