Skip to Content
Merck
All Photos(4)

Key Documents

373001

Sigma-Aldrich

Poly(ethylene glycol)

average Mn 4,600, hydroxyl

Synonym(s):

Polyethylene glycol, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Product Name

Poly(ethylene glycol), average Mn 4,600

form

flakes

mol wt

average Mn 4,400-4,800
average Mn 4,600

viscosity

180 cSt(210 °F)

mp

57-61 °C (lit.)

Ω-end

hydroxyl

α-end

hydroxyl

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (PEG) is a biocompatible water soluble poly(ether) with low toxicity. It is majorly used as a biopolymer for the surface modification of biomaterials. Its properties include hydrophilicity and non-immunogenicity.[1][2]

Application

PEG can be used in the formation of polymeric hydrogels for the formation of biomedical applications. [1][2][3][4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morphology of Photopolymerized End-linked Poly(ethylene glycol) Hydrogels by Small Angle X-ray Scattering
Waters DJ, et al.
Macromolecules, 43(16), 6861-6861 (2010)
Bacterial adhesion is affected by the thickness and stiffness of poly (ethylene glycol) hydrogels
Kolewe KW, et al.
ACS Applied Materials & Interfaces, 10(3), 2275-2281 (2018)
Nano-magnetic particles used in biomedicine: Core and coating materials
Karimi Z, et al.
Materials Science and Engineering Technology, 33(5), 2465-2475 (2013)
Biomedical hydrogels
Burdick JA, et al.
Biomaterials, Artificial Organs and Tissue Engineering, 33(5), 107-115 (2005)
Mark A Rice et al.
Acta biomaterialia, 5(1), 152-161 (2008-09-17)
Ultrasound has potential as a non-destructive analytical technique to provide real-time online assessments of matrix evolution in cell-hydrogel constructs used in tissue engineering. In these studies, chondrocytes were encapsulated in poly(ethylene glycol) hydrogels, and gel degradation was manipulated to provide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service