Skip to Content
Merck
All Photos(1)

Key Documents

93350

Sigma-Aldrich

Trizma® base

≥99.7% (T), crystalline, puriss p.a., aminopeptidase substrate

Synonym(s):

2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14
Beilstein:
741883
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Product Name

Trizma® base, puriss. p.a., ≥99.7% (T)

description

aminopeptidase substrate

Quality Level

grade

puriss. p.a.

Assay

≥99.7% (T)

form

crystalline

ign. residue

≤0.01% (as SO4)

pH

10.5-11.5 (25 °C, 10% in H2O)

useful pH range

7-9

pKa (25 °C)

8.1

bp

219-220 °C/10 mmHg (lit.)

mp

167-172 °C (lit.)
168-172 °C

solubility

water: soluble (678 g/l at 20 °C)

anion traces

chloride (Cl-): ≤5 mg/kg
sulfate (SO42-): ≤5 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

10 % in H2O

UV absorption

λ: 230 nm Amax: ≤0.6
λ: 260 nm Amax: ≤0.05
λ: 280 nm Amax: ≤0.04
λ: 430 nm Amax: ≤0.01

SMILES string

NC(CO)(CO)CO

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2

InChI key

LENZDBCJOHFCAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris is an established basimetric standard and buffer used in biochemistry and molecular biology.[1] It may be used by itself as a buffer or as a component of mixed buffer formulations, such as Tris-EDTA (TE) buffer, Tris-acetate-EDTA (TAE) buffer, Tris-borate-EDTA (TBE) buffer, etc.[2] It is pure, essentially stable, relatively non-hygroscopic and has a high equivalent weight.[3]

Application

Tris base has been used:
  • In the preparation of TAE buffer for TAE agarose gel used to run PCR products by gel electrophoresis[4]
  • As a component of sample buffer for SDS-PAGE[5]
  • In the preparation of IEC (ion-exchange chromatography) buffer[6]

Other Notes

Easily compare specifications for Trizma products with the Trizma specification table.
The pH values of all buffers are temperature- and concentration-dependent. For Tris buffers, pH increases about 0.03 unit per °C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution.
For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vivo Analysis of Cyclic di-GMP Cyclase and Phosphodiesterase Activity in Escherichia coli Using a Vc2 Riboswitch-based Assay.
Liu Y, et al.
Journal of Bacteriology (2017)
a-Synuclein Aggregation Monitored by Thioflavin T Fluorescence Assay.
Wordehoff M and Hoyer W
Bio-protocol, 8(14) (2018)
Quantitative determination of titin and nebulin in poultry meat by SDS-PAGE with an internal standard.
Tomaszewska-Gras J, et al.
Meat Science, 62(1), 61-66 (2002)
Preparation of Buffers for Use in Enzyme Studies.
Gomori G
Methods in Enzymology, 1, 138-146 (1955)
Sambrook J and Russell DW
Molecular Cloning: A Laboratory Manual (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service