Skip to Content
Merck
All Photos(1)

Key Documents

88417

Sigma-Aldrich

MTT Formazan

MTT Formazan

MTT reduction end product, powder

Synonym(s):

1-(4,5-Dimethylthiazol-2-yl)-3,5-diphenylformazan, Thiazolyl blue formazan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17N5S
CAS Number:
Molecular Weight:
335.43
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83
Pricing and availability is not currently available.

Product Name

MTT Formazan, MTT reduction end product

Quality Level

form

powder

solubility

chloroform: 10 mg/mL, clear, dark violet

SMILES string

Cc1nc(\N=N\C(=N\Nc2ccccc2)c3ccccc3)sc1C

InChI

1S/C18H17N5S/c1-13-14(2)24-18(19-13)23-22-17(15-9-5-3-6-10-15)21-20-16-11-7-4-8-12-16/h3-12,20H,1-2H3/b21-17+,23-22+

InChI key

GIFKVQOPAJDJLO-SZVLODPRSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

MTT Formazan has been used in cell viability assay.[1][2][3]

Biochem/physiol Actions

MTT assay is a colorimetric reaction, generally used to examine cell viability. It involves the enzymatic reduction of a yellow-colored, water-soluble, 3-[4,5-dimethylthiazole-2-yl]-2,5-diphenyltetrazolium bromide (MTT) to purple-colored, water-insoluble, MTT-formazan. This reaction is catalyzed by mitochondrial dehydrogenase and only occurs in viable cells.[4][5] Thus, the amount of blue formazan is proportional to the number of viable cells. The absorbance of the formazan solution is analyzed spectrophotometrically at 570 nm.[6]

Other Notes

Reviews: Tetrazolium salts and formazans[7]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymeric nanoparticles for the selective therapy of inflammatory bowel disease
Test, 508, 377-397 (2012)
Chao Zhang et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 80, 253-259 (2016-05-03)
Costunolide, an active sesquiterpene lactone, is derived from many herbal medicines and it exhibits a broad spectrum of bioactivities such as anti-inflammatory, potential anti-tumor activity. Herein we assessed the anti-cancer effects of costunolide on U2OS cells and explored the underlying
Jianing Li et al.
Pathology, research and practice, 215(10), 152591-152591 (2019-08-26)
The role of transforming growth factor beta (TGF-β) in lung cancer is well known. TGF-β-mediated cellular proliferation and angiogenesis through similar to mothers against decapentaplegic homolog 2 (Smad2) protein has also been well elucidated. Smad2 is a predicted target for
Nanotoxicology: evaluating toxicity potential of drug-nanoparticles
Nanoparticulate drug delivery, 1, 123-155 (2012)
Primary cultures of cardiac myocytes as in vitro models for pharmacological and toxicological assessments
In Vitro Methods in Pharmaceutical Research, 209-223 (1997)

Questions

Reviews

Active Filters

    • Review 1
    • Votes 8
    4 out of 5 stars.

    Will Not Work For MTT Proliferation Assay

    I accidentally bought this thinking it was MTT salt to use in a cell proliferation assay but it is MTT Formazan, the water insoluble end product! Therefore, it is useless for the assay. I suppose you could use it as a control to optimize your buffer for solubilizing the insoluble Formazan. The description for this product is misleading and should clarify that this is not what you need for an MTT assay, and refer the customer to MTT salt. That said, this is a nice purple crystalline powder and insoluble in water consistent with MTT Formazan, if that is what you actually want.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service