Skip to Content
Merck

M2003

MTT Formazan

powder

Synonym(s):

1-(4,5-Dimethylthiazol-2-yl)-3,5-diphenylformazan, Thiazolyl blue formazan

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H17N5S
CAS Number:
Molecular Weight:
335.43
NACRES:
NA.47
PubChem Substance ID:
UNSPSC Code:
12171500
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥97% (TLC)

Quality Segment

form

powder

color

purple

solubility

acetone: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cc1nc(\N=N\C(=N\Nc2ccccc2)c3ccccc3)sc1C

InChI

1S/C18H17N5S/c1-13-14(2)24-18(19-13)23-22-17(15-9-5-3-6-10-15)21-20-16-11-7-4-8-12-16/h3-12,20H,1-2H3/b21-17+,23-22+

InChI key

GIFKVQOPAJDJLO-SZVLODPRSA-N

Application

MTT Formazan has been used to measure cell viability.

Biochem/physiol Actions

MTT assay is a colorimetric reaction, generally used to examine cell viability. It involves the enzymatic reduction of a yellow-colored, water-soluble, 3-[4,5-dimethylthiazole-2-yl]-2,5-diphenyltetrazolium bromide (MTT) to purple-colored, water-insoluble, MTT-formazan. This reaction is catalyzed by mitochondrial dehydrogenase and only occurs in viable cells. Thus, the amount of blue formazan is proportional to the number of viable cells. The absorbance of the formazan solution is analyzed spectrophotometrically at 570 nm.


Still not finding the right product?

Explore all of our products under MTT Formazan


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library