Skip to Content
Merck
All Photos(1)

Key Documents

85590

Sigma-Aldrich

Spermine

≥99.0% (GC)

Synonym(s):

N,N′-Bis(3-aminopropyl)-1,4-diaminobutane, Gerontine, Musculamine, Neuridine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)3NH(CH2)4NH(CH2)3NH2
CAS Number:
Molecular Weight:
202.34
Beilstein:
1750791
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.25

biological source

microbial
synthetic

Quality Level

Assay

≥99.0% (GC)

form

crystalline (chunks)

impurities

≤0.5% spermidine

bp

150 °C/5 mmHg (lit.)

mp

28-30 (lit.)

solubility

water: 0.05 g/mL, clear, colorless

anion traces

carbonate (CO32-): ≤5000 mg/kg

storage temp.

2-8°C

SMILES string

[H]N(CCCN)CCCCN([H])CCCN

InChI

1S/C10H26N4/c11-5-3-9-13-7-1-2-8-14-10-4-6-12/h13-14H,1-12H2

InChI key

PFNFFQXMRSDOHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Spermine, a polycationic biogenic polyamine derived from spermidine, may be used in a wide variety of biological applications as a supplement or regulatory agent.Used as co-matrix with DHB for MALDI-MS of sialylated glycans in negative ion mode.

Biochem/physiol Actions

Mixed NMDA agonist/antagonist at the polyamine site. Plays a role in cellular proliferation and differentiation; inhibits neuronal nitric oxide synthase (nNOS).
Mixed NMDA glutamate receptor agonist/antagonist at the polyamine site. Neuroprotective effects have been observed at high concentrations (1 mM), while neurotoxicity is observed at lower concentrations. It enhances agonist effectiveness at the strychnine-insensitive glycine site. Plays a role in cellular proliferation and differentiation; inhibits neuronal nitric oxide synthase (nNOS).

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Mechref et al.
Journal of the American Society for Mass Spectrometry, 9(12), 1293-1302 (1998-12-03)
Negative-ion matrix-assisted laser desorption/ionization mass spectra of sialyated glycoconjugates were acquired employing 2,5-dihydroxybenzoic acid (DHB) in conjunction with spermine as a co-matrix. The addition of spermine to DHB permitted an improved crystal formation as well as a higher analyte solubility.
Nicole K L Lee et al.
Journal of cellular physiology, 226(6), 1453-1460 (2011-03-18)
The naturally occurring polyamines, spermidine, spermine, and their precursor putrescine, play indispensible roles in both prokaryotic and eukaryotic cells, from basic DNA synthesis to regulation of cell proliferation and differentiation. The rate-limiting polyamine biosynthetic enzymes, ornithine decarboxylase (ODC) and S-adenosylmethionine
Lydia P P Liew et al.
Bioorganic & medicinal chemistry letters, 23(2), 452-454 (2012-12-26)
Screening of synthesized and isolated marine natural products for in vitro activity against four parasitic protozoa has identified the ascidian metabolite 1,14-sperminedihomovanillamide (orthidine F, 1) as being a non-toxic, moderate growth inhibitor of Plasmodium falciparum (IC(50) 0.89 μM). Preliminary structure-activity
Brian R Lee et al.
Nature neuroscience, 16(11), 1644-1651 (2013-10-01)
In rat models of drug relapse and craving, cue-induced cocaine seeking progressively increases after withdrawal from the drug. This 'incubation of cocaine craving' is partially mediated by time-dependent adaptations at glutamatergic synapses in nucleus accumbens (NAc). However, the circuit-level adaptations
Wendy Wai Yeng Yeo et al.
Journal of nanoscience and nanotechnology, 14(5), 3328-3336 (2014-04-17)
A nano-sized polymer, dextran-spermine (D-SPM), was shown to have the capacity to deliver gene to the lung of mouse via intranasal route. In this study, assessments on the safety profile of D-SPM were performed to complement the gene expression results.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service