Skip to Content
Merck
All Photos(1)

Key Documents

80635

Sigma-Aldrich

PIPES

BioXtra, for molecular biology, ≥99.5% (T)

Synonym(s):

1,4-Piperazinediethanesulfonic acid, Piperazine-1,4-bis(2-ethanesulfonic acid), Piperazine-N,N′-bis(2-ethanesulfonic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N2O6S2
CAS Number:
Molecular Weight:
302.37
Beilstein:
817713
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

grade

for molecular biology

Quality Level

product line

BioXtra

Assay

≥99.5% (T)

form

powder

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

ign. residue

≤0.2% (as SO4)

loss

≤0.2% loss on drying, 110 °C

useful pH range

6.1-7.5

pKa (25 °C)

6.8

mp

>300 °C (lit.)

solubility

0.1 M NaOH: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤200 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.1 M in 1 M NaOH

UV absorption

λ: 260 nm Amax: 0.055
λ: 280 nm Amax: 0.040

SMILES string

OS(=O)(=O)CCN1CCN(CC1)CCS(O)(=O)=O

InChI

1S/C8H18N2O6S2/c11-17(12,13)7-5-9-1-2-10(4-3-9)6-8-18(14,15)16/h1-8H2,(H,11,12,13)(H,14,15,16)

InChI key

IHPYMWDTONKSCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Sensitive Detection of Immunoglobulin G Stability Using in Real-Time Isothermal Differential Scanning Fluorimetry: Determinants of Protein Stability for Antibody-Based Therapeutics.: The use of PIPES buffer in this study was integral to maintaining the stability of immunoglobulin G during differential scanning fluorimetry. This method is crucial for evaluating the stability of antibody-based therapeutics (Moggridge et al., 2017).

Other Notes

Hybrid selection of mRNA - component of prehybridization buffer

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Jagus
Methods in Enzymology, 152, 570-570 (1987)
Veronique Adoue et al.
Molecular systems biology, 10, 754-754 (2014-10-19)
Most complex disease-associated genetic variants are located in non-coding regions and are therefore thought to be regulatory in nature. Association mapping of differential allelic expression (AE) is a powerful method to identify SNPs with direct cis-regulatory impact (cis-rSNPs). We used
Nicholas Light et al.
Epigenetics, 9(9), 1238-1251 (2014-07-24)
Allele-specific (AS) assessment of chromatin has the potential to elucidate specific cis-regulatory mechanisms, which are predicted to underlie the majority of the known genetic associations to complex disease. However, development of chromatin landscapes at allelic resolution has been challenging since
J Mano et al.
Biochimica et biophysica acta, 1480(1-2), 374-376 (2000-09-27)
Arabidopsis thaliana P1 protein was crystallized using the hanging drop vapor-diffusion method in 0.1 M piperazine-1, 4-bis(2-ethanesulfonic acid) buffer, containing 14% polyethylene glycol 6000 and 0.2 M magnesium acetate at pH 6.5 and 20 degrees C. The crystals are orthorhombic
B W Nielsen et al.
Agents and actions, 35(3-4), 170-178 (1992-03-01)
Increased osmotic pressure has been reported to cause non-cytotoxic histamine release (HR) from human basophils, as well as a potentiation of HR induced by anti-IgE. In this study, the effects of hyperosmolar Na-K-acetate (300-600 mOsm/kg H2O) on HR was studied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service