Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
PIPES
Synonym(s):
1,4-Piperazinediethanesulfonic acid, Piperazine-1,4-bis(2-ethanesulfonic acid), Piperazine-N,N′-bis(2-ethanesulfonic acid)
Empirical Formula (Hill Notation):
C
8
H
18
N
2
O
6
S
2
CAS Number:
5625-37-6
Molecular Weight:
302.37
EC Number:
227-057-6
Products
Product Category
buffers (5)
Brand
Sigma-Aldrich (5)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Quality Segment
Application
Special Grade
Anhydrous (1)
BioPerformance Certified (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
P6757
PIPES,
≥99% (titration)
Expand
View Pricing
P1851
PIPES,
BioPerformance Certified, suitable for cell culture
Expand
View Pricing
80635
PIPES,
BioXtra, for molecular biology, ≥99.5% (T)
Expand
View Pricing
P8203
PIPES,
BioXtra, ≥99% (titration)
Expand
View Pricing
RDD004
PIPES,
anhydrous, free-flowing, Redi-Dri
™
, ≥99%
Expand
View Pricing