Skip to Content
Merck
All Photos(1)

Key Documents

79851

Sigma-Aldrich

Kaempferol 3-β-D-glucopyranoside

≥97.0% (HPLC)

Synonym(s):

3,4′,5,7-Tetrahydroxyflavone 3-glucoside, 3-(β-D-Glucopyranosyloxy)-5,7-dihydroxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-one, 3-Glucosylkaempferol, Astragalin, Kaempferol 3-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O11
CAS Number:
Molecular Weight:
448.38
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.47

Assay

≥97.0% (HPLC)

form

crystals

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)cc4)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C21H20O11/c22-7-13-15(26)17(28)18(29)21(31-13)32-20-16(27)14-11(25)5-10(24)6-12(14)30-19(20)8-1-3-9(23)4-2-8/h1-6,13,15,17-18,21-26,28-29H,7H2/t13-,15-,17+,18-,21+/m1/s1

InChI key

JPUKWEQWGBDDQB-QSOFNFLRSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Role in Ubiquinone Biosynthesis: The compound contributed to the biosynthesis of the benzenoid moiety of ubiquinone, a key component in plant biochemistry, highlighting its essential role in cellular energy production and plant health (Soubeyrand et al., 2018).

Biochem/physiol Actions

Similar kaempferol glycosides reduce food intake and corresponding weight gain in Wistar rats. The glucoside was not tested.
Similar kaempferol glycosides reduce food intake and corresponding weight gain in Wistar rats.The glucoside was not tested.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingsheng Peng et al.
Journal of experimental botany, 59(11), 2933-2944 (2008-06-17)
Plants can survive a limiting nitrogen (N) supply by developing a set of N limitation adaptive responses. However, the Arabidopsis nla (nitrogen limitation adaptation) mutant fails to produce such responses, and cannot adapt to N limitation. In this study, the
Xianghui He et al.
Planta medica, 77(17), 1950-1957 (2011-06-16)
Cuscuta chinensis and Cuscuta australis, the two botanical sources of the Chinese herbal medicine Tu-Si-Zi, were distinguished from each other based on qualitative and quantitative chemical analysis. By HPLC‑DAD‑MS, a total of 36 compounds were characterized from these two Cuscuta
Hyang-Bok Lee et al.
Journal of the science of food and agriculture, 91(13), 2315-2321 (2011-05-14)
The application of tea seed extract (TSE) has been widely investigated because of its biological activities. In this paper, two flavonol triglycosides in TSE-camelliaside A (CamA) and camelliaside B (CamB)-were subjected to hydrolysis in the presence of two commercial enzyme
Min Ke et al.
Bio-medical materials and engineering, 22(1-3), 113-119 (2012-07-07)
Diabetic retinopathy (DR) is a severe complication of diabetes mellitus (DM) and often causes vision loss or even blindness. Vascular endothelial growth factor (VEGF) in the retina, which is mainly derived from Müller cells, is a crucial biological factor in
Jiachen Zi et al.
Journal of bioscience and bioengineering, 112(4), 360-362 (2011-07-12)
Incubation of quercetin with Cunninghamella elegans ATCC 9245 yielded three metabolites, including quercetin 3-O-β-D-glucopyranoside, kaempferol 3-O-β-D-glucopyranoside and isorhamnetin 3-O-β-D-glucopyranoside. Glucosylation, O-methylation and dehydroxylation were involved in the process, among which dehydroxylation has never been found in Cunninghamella. Quercetin was completely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service