Skip to Content
Merck
All Photos(1)

Key Documents

77607

Sigma-Aldrich

Phenol

BioUltra, for molecular biology, TE-saturated, ~73% (T)

Synonym(s):

Hydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OH
CAS Number:
Molecular Weight:
94.11
Beilstein:
969616
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.25
grade:
Molecular Biology
for molecular biology
bp:
182 °C (lit.)
vapor pressure:
0.09 psi ( 55 °C)
0.36 mmHg ( 20 °C)

grade

Molecular Biology
for molecular biology

Quality Level

vapor density

3.24 (vs air)

vapor pressure

0.09 psi ( 55 °C)
0.36 mmHg ( 20 °C)

product line

BioUltra

Assay

~73% (T)

form

liquid

autoignition temp.

1319 °F

quality

TE-saturated

contains

~0.1% 8-hydroxyquinoline as preservative

expl. lim.

8.6 %

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

refractive index

n20/D 1.48

pH

~7.8(aqueous phase)

bp

182 °C (lit.)

mp

40-42 °C (lit.)

density

1.071 g/mL at 25 °C (lit.)

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

Oc1ccccc1

suitability

suitable for molecular biology

storage temp.

2-8°C

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

General description

Phenol, also known as hydroxybenzene, is an aromatic compound produced both naturally and synthetically. It acts as a disinfectant and is highly toxic. Exposure to phenol through inhalation, cutaneous adsorption, or oral ingestion causes denaturation and precipitation of cellular protein, which in turn leads to poisoning.
saturated with 10 mM TRIS buffer pH 8.0 and 1 mM EDTA

Application

Phenol (TE-saturated) has been used in the phenol-chloroform extraction procedure for plasmid DNA isolation. It has also been used to extract the aqueous phase during DNA linearization and mRNA synthesis.

Caution

according to temperature, a small quantity of an overlaid aqueous phase may form

Other Notes

RNA fingerprinting - preparation of the E. coli tRNA carrier

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biodegradation of phenol: mechanisms and applications
van Schie PM and Young LY
Bioremediation Journal, 1222-1224 (2000)
Phenol and its toxicity: A case report
Chand M, et al.
Iranian Journal of Toxicology, 247-274 (2015)
Induction of Natural Competence in Genetically-modified Lactococcus lactis
Mulder J, et al.
Bio-protocol, 1-18 (2018)
A.D. Branch et al.
Methods in Enzymology, 180, 133-133 (1989)
Mannosylated and histidylated LPR technology for vaccination with tumor antigen mRNA
Pichon C and Midoux P
Methods in Molecular Biology (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service