Skip to Content
Merck
All Photos(1)

Key Documents

72069

Sigma-Aldrich

Sodium tungstate dihydrate

BioUltra, ≥99.0% (T)

Synonym(s):

Tungstic acid sodium salt dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2WO4 · 2H2O
CAS Number:
Molecular Weight:
329.85
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25
Assay:
≥99.0% (T)
form:
crystals
solubility:
H2O: 1 M at 20 °C, clear, colorless

product line

BioUltra

Quality Level

Assay

≥99.0% (T)

form

crystals

impurities

insoluble matter, passes filter test
≤0.001% total nitrogen (N)
≤0.1% free alkali (as NaOH)

pH

9.5-11.5 (25 °C, 1 M in H2O)

mp

698 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

As: ≤0.5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤10 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: ≤0.120
λ: 280 nm Amax: ≤0.035

SMILES string

O.O.[Na+].[Na+].[O-][W]([O-])(=O)=O

InChI

1S/2Na.2H2O.4O.W/h;;2*1H2;;;;;/q2*+1;;;;;2*-1;

InChI key

WPZFLQRLSGVIAA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium tungstate dihydrate is a sodium salt of tungstic acid, which occurs as white colorless crystals.

Application

Sodium tungstate dihydrate has been used in bioluminescence assays and to synthesize polycrystalline samples.

Other Notes

Stain in electron microscopy (nucleic acids, certain glycoproteins); Protein precipitant.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural, vibrational and luminescence properties of the (1- x) CaWO4- xCdWO4 system
Taoufyq A, et al.
Journal of Solid State Chemistry, 219, 127-137 (2014)
R.M.C. Dawson, et al.
Data for Biochemical Research, 549-549 (1986)
Iron is involved in maintenance of circadian period length in Arabidopsis
Chen YY, et al.
Plant Physiology, pp-112 (2013)
J.C. Stockert
Biology of the Cell, 29, 211-211 (1977)
Gardner's Commercially Important Chemicals: Synonyms, Trade Names, and Properties (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service