Skip to Content
Merck
All Photos(3)

Key Documents

379751

Sigma-Aldrich

Sodium tungstate dihydrate

99.995% trace metals basis

Synonym(s):

Tungstic acid sodium salt dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2WO4 · 2H2O
CAS Number:
Molecular Weight:
329.85
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.995% trace metals basis

form

chunks

mp

698 °C (lit.)

SMILES string

O.O.[Na+].[Na+].[O-][W]([O-])(=O)=O

InChI

1S/2Na.2H2O.4O.W/h;;2*1H2;;;;;/q2*+1;;;;;2*-1;

InChI key

WPZFLQRLSGVIAA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José Angel Martín Rodriguez et al.
Journal of plant physiology, 167(8), 606-613 (2010-01-19)
We examined whether the reduced mycorrhization of abscisic acid (ABA)-deficient tomato mutants correlates with their incapacity in ABA biosynthesis and whether this effect is dependent on ethylene production. The mycorrhization of notabilis and sitiens mutants, which have different ABA deficiencies
Marta Amigó-Correig et al.
PloS one, 7(7), e39087-e39087 (2012-07-18)
This study aims at exploring the effects of sodium tungstate treatment on hypothalamic plasticity, which is known to have an important role in the control of energy metabolism. Adult lean and high-fat diet-induced obese mice were orally treated with sodium
Josie E Bamford et al.
Environmental toxicology and chemistry, 30(10), 2312-2318 (2011-08-02)
Due to unknown effects of the potential exposure of the terrestrial environment to tungsten substances, a series of toxicity studies of sodium tungstate (Na(2) WO(4) ) was conducted. The effect on earthworm (Eisenia fetida) survival and reproduction was examined using
S M McInturf et al.
Toxicology and applied pharmacology, 254(2), 133-137 (2011-02-08)
The debate on tungsten (W) is fostered by its continuous usage in military munitions. Reports demonstrate W solubilizes in soil and can migrate into drinking water supplies and, therefore, is a potential health risk to humans. This study evaluated the
C J Rodriguez-Hernandez et al.
FEBS letters, 587(10), 1579-1586 (2013-04-17)
Both radiotherapy and most effective chemotherapeutic agents induce different types of DNA damage. Here we show that tungstate modulates cell response to DNA damaging agents. Cells treated with tungstate were more sensitive to etoposide, phleomycin and ionizing radiation (IR), all

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service