Skip to Content
Merck
All Photos(1)

Key Documents

40165

Sigma-Aldrich

Dimethyldioctadecylammonium bromide

≥98.0% (AT)

Synonym(s):

Distearyldimethylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)17]2N(Br)(CH3)2
CAS Number:
Molecular Weight:
630.95
Beilstein:
3580323
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

cationic

Quality Level

Assay

≥98.0% (AT)

mol wt

630.95 g/mol

technique(s)

dialysis: suitable
enzyme immunoassay: suitable
protein purification: suitable

mp

~160 °C

SMILES string

CCCCCCCCCCCCCCCCCC[N+](CCCCCCCCCCCCCCCCCC)(C)C.[Br-]

InChI

1S/C38H80N.BrH/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-39(3,4)38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-2;/h5-38H2,1-4H3;1H/q+1;/p-1

InChI key

PSLWZOIUBRXAQW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keith M Hamel et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(1), 60-67 (2014-01-23)
Inducible costimulator (ICOS)-ICOSL interactions are necessary for activation of Teff cells and follicular helper T (Tfh) cells. ICOSL is expressed on B cells, macrophages, and dendritic cells and can be induced on nonhematopoietic cells. The aim of this study was
Paolo Sabatino et al.
The journal of physical chemistry. B, 116(30), 9269-9276 (2012-06-27)
The interaction between phenol molecules, both in their undissociated and dissociated states, and cationic dioctadecyl dimethylammonium chloride (DODAC) vesicles were thoroughly investigated using NMR techniques. In particular, diffusion and relaxation measurements, combined with the two sites Kärger model, were used
Zhi Rao et al.
Colloids and surfaces. B, Biointerfaces, 97, 248-253 (2012-05-23)
In order to confirm that the cholesterol end groups of cholesterol-end capped polyethylene glycol really insert into the liposome bilayer and investigate how the incorporation affects the microenvironment of liposome bilayer, two kinds of molecular probes, namely Nile Red and
A P Dabkowska et al.
Biomacromolecules, 13(8), 2391-2401 (2012-06-26)
The interaction of DNA with monolayers of the cationic lipid dimethyldioctadecylammonium bromide, with/without 50 mol % of a neutral "helper" lipid, either dioleoylphosphatidylethanolamine or cholesterol, has been studied using specular neutron reflection, surface pressure-area isotherms, and Brewster angle microscopy. The
Randip Kaur et al.
Journal of controlled release : official journal of the Controlled Release Society, 164(3), 331-337 (2012-07-18)
The mechanism behind the immunostimulatory effect of the cationic liposomal vaccine adjuvant dimethyldioctadecylammonium and trehalose 6,6'-dibehenate (DDA:TDB) has been linked to the ability of these cationic vesicles to promote a depot after administration, with the liposomal adjuvant and the antigen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service