Skip to Content
Merck
All Photos(1)

Key Documents

330M-1

Sigma-Aldrich

S-100 (4C4.9) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

diluted ascites fluid

antibody product type

primary antibodies

clone

4C4.9, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (330M-14)
vial of 0.5 mL concentrate (330M-15)
bottle of 1.0 mL predilute (330M-17)
vial of 1.0 mL concentrate (330M-16)
bottle of 7.0 mL predilute (330M-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:200

isotype

IgG2a

control

melanoma

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic, nuclear

Gene Information

human ... S100A1(6271)

General description

S-100 protein has been found in normal melanocytes, Langerhans cells, histiocytes, chondrocytes, lipocytes, skeletal and cardiac muscle, Schwann cells, epithelial and myoepithelial cells of the breast, salivary and sweat glands, as well as in glial cells.[1][2][3]Neoplasms derived from these cells also express S-100 protein, albeit non-uniformly.[1][2][4][5] A large number of well differentiated tumors of the salivary gland, adipose and cartilaginous tissue,[4] and Schwann cell-derived tumors express S-100 protein. Almost all malignant melanomas and cases of histiocytosis X are positive for S-100 protein.[5][6] Despite the fact that S-100 protein is an ubiquitous substance, its demonstration is of great value in the identification of several neoplasms, particularly melanomas.[1][2][4][5][6][3]

Quality


IVD

IVD

IVD

RUO

Linkage

S-100 Positive Control Slides, Product No. 330S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Monda et al.
Human pathology, 16(3), 287-293 (1985-03-01)
In an effort to investigate the utility of immunostaining for S-100 protein in the differential diagnosis of chondroblastoma, the expression of S-100 protein in nine chondroblastomas was compared with that in six giant cell tumors, six aneurysmal bone cysts, four
Purvisha Patel et al.
Journal of the American Academy of Dermatology, 46(2), 264-270 (2002-01-25)
Malignant myxoid melanoma (MMM) is a rarely reported variant of malignant melanoma, which can often be confused with other mucin-containing neoplasms. A retrospective study of 3 cases of MMM and a review of the English-language literature was performed. MMM affects
K M McLaren et al.
Human pathology, 27(7), 633-636 (1996-07-01)
In general, the diagnosis of papillary carcinoma of the thyroid is readily achieved based on a defined aggregate of histopathologic features. A papillary architecture is an important but not pivotal component of the diagnosis. The recognition of classic nuclear features
T Nakajima et al.
The American journal of surgical pathology, 6(8), 715-727 (1982-12-01)
The presence of S-100 protein was immunohistochemically studied in many types of formalin-fixed and paraffin-embedded tumors (260 cases). Peripheral nerve tumors, i.e., schwannomas, neurofibromas, granular cell tumors, and neurogenic sarcomas were demonstrated to contain variable amounts of S-100 protein in
H J Kahn et al.
American journal of clinical pathology, 79(3), 341-347 (1983-03-01)
Normal tissues and various tumors were examined for S100 protein, using anti-S100 protein antiserum, in an immunoperoxidase reaction. Among normal tissues, in addition to the previously reported presence of S100 protein in some neurons, glial, and Schwann cells of the

Articles

IHC antibodies enhance dermatopathology beyond H&E stained slides, improving techniques and applications for dermatological research.

IHC antibodies enhance dermatopathology beyond H&E stained slides, improving techniques and applications for dermatological research.

IHC antibodies enhance dermatopathology beyond H&E stained slides, improving techniques and applications for dermatological research.

IHC antibodies enhance dermatopathology beyond H&E stained slides, improving techniques and applications for dermatological research.

Questions

  1. What are the subunits for S100 (4C4.9), 330M-18?

    1 answer
    1. The S100 protein 330M-18 consists of both alpha and beta subunits. This protein belongs to the family of calcium-binding proteins, similar to calmodulin and troponin C. Specifically, S100A is composed of an alpha and beta chain, while S100B is composed of two beta chains. Furthermore, the S100 protein is expressed in antigen-presenting cells, including Langerhans cells in the skin and interdigitating reticulum cells in the paracortex of lymph nodes.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service