Skip to Content
Merck
All Photos(1)

Key Documents

343M-9

Sigma-Aldrich

TTF-1 (8G7G3/1) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

8G7G3/1, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (343M-94)
vial of 0.5 mL concentrate (343M-95)
bottle of 1.0 mL predilute (343M-97)
vial of 1.0 mL concentrate (343M-96)
bottle of 7.0 mL predilute (343M-98)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1

control

lung adenocarcinoma

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... A1CF(29974)

General description

Anti-TTF-1 (Thyroid Transcription Factor 1) is useful in differentiating primary adenocarcinoma of the lung from metastatic carcinomas originating in the organs rather than thyroid, germ cell tumors, and malignant mesothelioma. It can also be used to differentiate small cell lung carcinoma from lymphoid infiltrates. TTF-1 labeling is also seen in thyroid and thyroid-derived tumors TTF-1 immunostaining is useful in the differentiation between pulmonary and nonpulmonary origin of adenocarcinomas in malignant effusions. TTF-1 staining is very reliable in discerning whether a brain metastasis has arisen from a pulmonary or nonpulmonary site, particularly when dealing with adenocarcinomas and large-cell carcinomas.
Thyroid transcription factor-1 (TTF-1) is a transcription factor that plays a role in regulating genes expressed within the thyroid, lungs, and brain. TTF-1 expression is present in normal thyroid epithelium and type II pneumocytes and nonciliated bronchiolar secretory cells of the lung. TTF-1 expression is seen in most primary lung and thyroid carcinomas, but rarely in other carcinomas. Anti-TTF-1 immunohistochemistry is useful for identifying carcinomas of thyroid and lung origin.

Quality


IVD

IVD

IVD

RUO

Linkage

TTF-1 Positive Control Slides, Product No. 343S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Di Loreto et al.
Journal of clinical pathology, 50(1), 30-32 (1997-01-01)
To investigate the immunocytochemical expression of the tissue specific transcription factor-1 (TTF-1) on cytological specimens of small cell lung carcinoma (SCLC) and to establish its value in the cytological diagnosis of lung cancer. For each case, the diagnosis was made
R Katoh et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 13(5), 570-576 (2000-05-29)
Thyroid transcription factor-1 (TTF-1) has been known to regulate the transcriptional activity of thyroid-specific genes. We examined the expression of TTF-1 in non-neoplastic and neoplastic thyroid tissues. By immunohistochemistry, the nuclei of normal and hyperplastic follicular cells strongly reacted with
Dongfeng Tan et al.
Human pathology, 34(6), 597-604 (2003-06-27)
Thyroid transcription factor 1 (TTF-1), a homeodomain-containing transcription factor, plays a pivotal role in lung development, cell growth, and differentiation processes. The current literature reports considerable variation in frequency of TTF-1 protein expression in human non-small cell lung cancer (NSCLC).
P A Bejarano et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 9(4), 445-452 (1996-04-01)
Antibodies to the pulmonary epithelial cell-specific proteins surfactant proteins A and B (SP-A and SP-B) and to thyroid transcription factor-1 (TTF-1), a homeodomain nuclear transcription protein, were used as immunohistochemical markers to asses their ability to distinguish primary pulmonary non-small
C Di Loreto et al.
Cancer letters, 124(1), 73-78 (1998-03-21)
TTF-1 is a tissue-specific transcription factor expressed in the epithelial cells of thyroid and lung. This study investigates the immunohistochemical expression of TTF-1 in pleural malignant mesotheliomas (MM) and adenocarcinomas (AC) of the lung, respectively. For this purpose, 33 biopsy

Articles

Colorectal cancer is the third most common cancer in both men and women. An estimated 136,000 cases of colorectal cancer are expected to occur in 2016.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service